Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00426283_m1
          See other HBD GE Assays ›
          Gene Symbol:
          HBD
          Gene Name:
          hemoglobin subunit delta
          Gene Aliases:
          HBK
          Chromosome Location:
          Chr.11: 5232838 - 5234483 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00426283_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HBD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000519.4 NP_000510.1 2 - 3 363 131
          GenBank mRNA AY034468.1 - 2 - 3 508 131
          BC069307.1 - 2 - 3 363 131
          BC070282.1 - 2 - 3 364 131
          BU664913.1 - 2 - 3 327 131

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100009918.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11001331.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HBD

          Gene Ontology Categories:

          Function(s) Process(es)

          carbon dioxide transport
          oxygen transport
          erythrocyte development
          oxygen transporter activity
          protein binding
          oxygen binding
          heme binding
          hemoglobin alpha binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11016755.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016755.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016755.hg.1 TC11001331.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11016751.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016751.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016751.hg.1 TC11001331.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016756.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016756.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016756.hg.1 TC11001331.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11016754.hg.1 ENST00000292901.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016754.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016754.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016754.hg.1 TC11001331.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016753.hg.1 ENST00000292901.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016753.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11016753.hg.1 NM_000519.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11016753.hg.1 TC11001331.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174862.hg.1 TC1100009918.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100174858.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174858.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174858.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174858.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174858.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174858.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174858.hg.1 TC1100009918.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174861.hg.1 TC1100009918.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100174863.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174863.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174863.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174863.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174863.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174863.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174863.hg.1 TC1100009918.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000335295.4 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000380315.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000475226.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000485743.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 ENST00000633227.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009917.hg.1 NM_000518.4 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000417377.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay TC1100009918.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 ENST00000292901.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 ENST00000429817.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174860.hg.1 TC1100009918.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100174864.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174864.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174864.hg.1 ENST00000380299.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100174864.hg.1 NM_000519.3 Refseq
          Affymetrix Human Clariom D Assay PSR1100174864.hg.1 TC1100009918.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline