Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00426586_m1
          See other TYMS GE Assays ›
          Gene Symbol:
          TYMS
          Gene Name:
          thymidylate synthetase
          Gene Aliases:
          DKCD, HST422, TMS, TS
          Chromosome Location:
          Chr.18: 657653 - 673578 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00426586_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TYMS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001071.4 NP_001062.1 1 - 2 293 60
          NM_001354867.2 NP_001341796.1 1 - 2 293 60
          GenBank mRNA AB062290.1 - 2 - 3 284 60
          AB077208.1 - 1 - 2 203 60
          AK223372.1 - 1 - 2 330 60
          AK292620.1 - 1 - 2 237 60
          AK314140.1 - 1 - 2 301 60
          BC002567.2 - 1 - 2 271 60
          BC013919.1 - 2 - 3 298 60
          BC018858.2 - 1 - 2 255 60
          BC083512.1 - 2 - 3 309 60
          BT006811.1 - 1 - 2 203 60
          HY016886.1 - 1 - 2 365 60
          X02308.1 - 1 - 2 308 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800006448.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC18000006.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TYMS

          Gene Ontology Categories:

          Function(s) Process(es)

          dTMP biosynthetic process
          dTTP biosynthetic process
          nucleotide biosynthetic process
          negative regulation of translation
          methylation
          tetrahydrofolate interconversion
          tetrahydrofolate metabolic process
          DNA biosynthetic process
          translation repressor activity, nucleic acid binding
          thymidylate synthase activity
          folic acid binding
          methyltransferase activity
          transferase activity
          transferase activity, transferring one-carbon groups
          sequence-specific mRNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR18000055.hg.1 ENST00000323274.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000055.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18000055.hg.1 TC18000006.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18000052.hg.1 ENST00000323274.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000052.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18000052.hg.1 TC18000006.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000584122.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 ENST00000584122.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010488.hg.1 ENST00000589501.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000057.hg.1 ENST00000323224.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000057.hg.1 ENST00000323274.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000057.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18000057.hg.1 TC18000006.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR18000056.hg.1 ENST00000323224.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000056.hg.1 ENST00000323250.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000056.hg.1 ENST00000323274.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR18000056.hg.1 NM_001071.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR18000056.hg.1 TC18000006.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1900010488.hg.1 ENST00000589501.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144204.hg.1 TC1800006448.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800144203.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144203.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144203.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144203.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144203.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144203.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144203.hg.1 TC1800006448.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323250.9 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000581920.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000584122.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 ENST00000584122.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay TC1800006448.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144199.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144199.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144199.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144199.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144199.hg.1 TC1800006448.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000584122.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000323224.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000323274.14 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000579128.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 ENST00000584122.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 NM_001071.2 Refseq
          Affymetrix Human Clariom D Assay PSR1800144206.hg.1 TC1800006448.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline