Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00428488_g1
          See other PRDX2 GE Assays ›
          Gene Symbol:
          PRDX2
          Gene Name:
          peroxiredoxin 2
          Gene Aliases:
          HEL-S-2a, NKEF-B, NKEFB, PRP, PRX2, PRXII, PTX1, TDPX1, TPX1, TSA
          Chromosome Location:
          Chr.19: 12796823 - 12801800 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00428488_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PRDX2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005809.6 NP_005800.3 1 - 2 61 160
          GenBank mRNA AK289485.1 - 1 - 2 91 160
          AK293957.1 - 1 - 2 91 160
          AL600939.1 - 1 - 2 61 160
          BC000452.2 - 1 - 2 50 160
          BC003022.1 - 1 - 2 50 160
          BC039428.1 - 1 - 2 110 160
          BC064138.1 - 1 - 2 50 160
          BG705975.1 - 1 - 2 145 160
          EU668325.1 - 1 - 2 50 160
          L19185.1 - 1 - 2 115 160
          Z22548.1 - 1 - 2 80 160

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900009752.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001210.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC000452.2, BC003022.1, BC039428.1, NM_005809.6, Z22548.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PRDX2

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to tumor cell
          respiratory burst involved in inflammatory response
          response to oxidative stress
          regulation of hydrogen peroxide metabolic process
          removal of superoxide radicals
          positive regulation of blood coagulation
          negative regulation of lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          cellular response to oxidative stress
          T cell proliferation
          hydrogen peroxide metabolic process
          hydrogen peroxide catabolic process
          regulation of apoptotic process
          T cell homeostasis
          negative regulation of apoptotic process
          positive regulation of MAPK cascade
          leukocyte activation
          cell redox homeostasis
          negative regulation of T cell differentiation
          thymus development
          extrinsic apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          peroxidase activity
          protein binding
          thioredoxin peroxidase activity
          antioxidant activity
          oxidoreductase activity
          peroxiredoxin activity
          thioredoxin-dependent peroxiredoxin activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900007097.hg.1 ENST00000585496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900007097.hg.1 ENST00000585496.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600008917.hg.1 ENST00000447134.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006757.hg.1 ENST00000450181.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500009692.hg.1 ENST00000515779.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017013.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017013.hg.1 ENST00000334482.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017013.hg.1 ENST00000435703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017013.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19017013.hg.1 TC19001210.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19017014.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017014.hg.1 ENST00000334482.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017014.hg.1 ENST00000435703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017014.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19017014.hg.1 TC19001210.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19017017.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017017.hg.1 ENST00000435703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19017017.hg.1 NM_005809.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19017017.hg.1 TC19001210.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000007270.hg.1 ENST00000451920.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007270.hg.1 ENST00000451920.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007270.hg.1 ENST00000451920.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179651.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179651.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179651.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179651.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179651.hg.1 TC1900009752.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1300006757.hg.1 ENST00000450181.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007270.hg.1 ENST00000451920.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000007270.hg.1 ENST00000451920.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000007270.hg.1 ENST00000451920.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179654.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179654.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179654.hg.1 TC1900009752.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500009692.hg.1 ENST00000515779.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000478908.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 ENST00000498785.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900009752.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay TC1900007097.hg.1 ENST00000585496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900007097.hg.1 ENST00000585496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179646.hg.1 TC1900009752.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000301522.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000334482.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000466174.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 ENST00000477555.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 NM_005809.5 Refseq
          Affymetrix Human Clariom D Assay PSR1900179648.hg.1 TC1900009752.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600008917.hg.1 ENST00000447134.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 158513 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline