Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00534343_s1
          See other MAFB GE Assays ›
          Gene Symbol:
          MAFB
          Gene Name:
          MAF bZIP transcription factor B
          Gene Aliases:
          KRML, MCTO
          Chromosome Location:
          Chr.20: 40685848 - 40689240 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00534343_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MAFB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_005461.4 NP_005452.2 1 1825 86
          GenBank mRNA AK027324.1 - 2 1364 86
          BC028098.1 - 1 1456 5242606 86
          BC036689.1 - 1 1826 5261053 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000009116.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000844.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MAFB

          Panther Classification:

          Molecular Function -

          basic leucine zipper transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          segment specification
          nervous system development
          sensory organ development
          respiratory gaseous exchange
          rhombomere 5 development
          rhombomere 6 development
          T cell differentiation in thymus
          brain segmentation
          inner ear morphogenesis
          negative regulation of erythrocyte differentiation
          negative regulation of osteoclast differentiation
          positive regulation of transcription from RNA polymerase II promoter
          thymus development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          protein binding
          transcription factor binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR20011571.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011571.hg.1 ENST00000396967.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011571.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011571.hg.1 TC20000844.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011575.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011575.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011575.hg.1 TC20000844.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20011573.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011573.hg.1 ENST00000396967.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011573.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011573.hg.1 TC20000844.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20011572.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20011572.hg.1 NM_005461.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011572.hg.1 TC20000844.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR2000163827.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163827.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000163827.hg.1 TC2000009116.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000163827.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163827.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000163826.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163826.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000163826.hg.1 TC2000009116.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000163826.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163826.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000163829.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163829.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay PSR2000163829.hg.1 TC2000009116.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000163829.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163829.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 ENST00000373313.2 Ensembl
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 NM_005461.4 Refseq
          Affymetrix Human Clariom D Assay TC2000009116.hg.1 NM_005461.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 226336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline