Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00536270_s1
          See other DLGAP1-AS2 GE Assays ›
          Gene Symbol:
          DLGAP1-AS2
          Gene Name:
          DLGAP1 antisense RNA 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 3603737 - 3610089 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00536270_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DLGAP1-AS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_119377.1 1 107 71
          GenBank mRNA AK312196.1 - 1 200 71
          BC094703.1 - 1 106 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1800006519.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3774867 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P164576 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 33967 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21818 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3774867 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P164576 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P164576 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 223974_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1560019_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 NR_119377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 NR_119377.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1800006519.hg.1 NR_119377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800144704.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144704.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144704.hg.1 NR_119377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800144704.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144704.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1800144704.hg.1 NR_119377.1 Refseq
          Affymetrix Human Clariom D Assay PSR1800144704.hg.1 TC1800006519.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000572856.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 ENST00000573177.1 Ensembl
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 NR_119377.1 Refseq
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 NR_119377.1 Refseq
          Affymetrix Human Clariom D Assay TC1800006519.hg.1 NR_119377.1 Refseq
          Human Genome U133 Plus 2.0 Array 223974_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560019_at - -
          Human Genome U133 Plus 2.0 Array 1560019_PM_at - -
          Human Genome U133 Plus 2.0 Array 223974_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560019_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 223974_at10 - -
          Human Genome U133 Set 223974_at - -
          Human Genome U133 Set 223974_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 161697 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3311145 - -
          HumanHT-12 v4 Expression BeadChip ILMN_3307982 BC005130.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3307982 BC094703.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3311160 BC005130.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline