Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00536733_s1
          See other NCBP2-AS2 GE Assays ›
          Gene Symbol:
          NCBP2-AS2
          Gene Name:
          NCBP2 antisense RNA 2 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 196942623 - 196944013 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00536733_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NCBP2-AS2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024388.1 1 521 69
          GenBank mRNA AK123020.1 - 1 470 69
          AL832915.1 - 1 457 69
          BC000257.1 - 1 453 69
          BC007882.2 - 1 432 69
          BC062368.1 - 1 521 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300010040.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001104.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 142513 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 57040 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 537513 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 AB029032.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 AK025057.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 AK095216.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 AK225886.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 AK309063.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 AL137384.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 CR936613.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 DQ335469.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 129268 NM_015312.2 refseq_mrna_human_NM
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P6708 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P80406 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P6708 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11638 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 19361 - -
          Agilent Human Genome CGH Microarray 44B A_14_P124917 - -
          Agilent Human Genome CGH Microarray 44A A_14_P124917 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 12001 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P6708 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P6708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23023272 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23023280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23023280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23023272 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02880808 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P6708 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P80406 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P124917 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P124917 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P124917 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P80406 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P6708 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 94970 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 99730 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P124917 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02880816 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02880811 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02880808 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02880814 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02880809 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P124917 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8084983 - -
          Affymetrix Human Gene 1.0 ST Array 8084984 - -
          Affymetrix Human Gene 1.0 ST Array 8084985 - -
          Affymetrix Human Gene 1.1 ST Array 8084982 - -
          Affymetrix HT HG-U133+ PM Array Plate 225657_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR03019060.hg.1 TC03001104.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019060.hg.1 NR_024388.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010040.hg.1 ENST00000602845.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010040.hg.1 NR_024388.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010040.hg.1 NR_024388.1 Refseq
          Affymetrix Human Clariom D Assay TC0300010040.hg.1 ENST00000602845.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010040.hg.1 NR_024388.1 Refseq
          Affymetrix Human Clariom D Assay TC0300010040.hg.1 NR_024388.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173133.hg.1 ENST00000602845.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173133.hg.1 NR_024388.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173133.hg.1 ENST00000602845.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173133.hg.1 NR_024388.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173133.hg.1 TC0300010040.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 225657_PM_at - -
          Human Genome U133 Plus 2.0 Array 225657_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 225657_at8 - -
          Human Genome U95 Set 59411_at - -
          Human Genome U95 Set 59411_PM_at - -
          Human Genome U133 Set 225657_at - -
          Human Genome U133 Set 225657_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023780 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023780 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 104483 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3248614 NR_024388.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3248614 AK123020.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248614 NR_024388.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248614 AK123020.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248614 NR_024388.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248614 AK123020.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248614 NR_024388.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248614 AK123020.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline