| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005616.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005616.hg.1 |
ENST00000381853.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005616.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005616.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005616.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005616.hg.1 |
TC13000496.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005617.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005617.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005617.hg.1 |
TC13000496.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005614.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005614.hg.1 |
ENST00000381853.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005614.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005614.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005614.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005614.hg.1 |
TC13000496.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005618.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005618.hg.1 |
ENST00000381853.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005618.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005618.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005618.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13005618.hg.1 |
TC13000496.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008381.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156582.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156582.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156582.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156582.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156582.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156582.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156582.hg.1 |
TC1300008381.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300156577.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156577.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156577.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156577.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156577.hg.1 |
TC1300008381.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156578.hg.1 |
TC1300008381.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
ENST00000357816.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
NM_199138.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156579.hg.1 |
TC1300008381.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300156580.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156580.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156580.hg.1 |
ENST00000515384.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300156580.hg.1 |
NM_152704.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300156580.hg.1 |
TC1300008381.hg.1 |
Clariom D TC |