Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00540173_s1
          See other KRTAP3-1 GE Assays ›
          Gene Symbol:
          KRTAP3-1
          Gene Name:
          keratin associated protein 3-1
          Gene Aliases:
          KAP3.1, KRTAP3.1
          Chromosome Location:
          Chr.17: 41008521 - 41009114 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00540173_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRTAP3-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031958.1 NP_114164.1 1 245 78
          GenBank mRNA AJ406931.1 - 1 245 78
          BC113077.1 - 1 225 40069149 78
          BC113078.1 - 1 229 40069150 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010647.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001478.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KRTAP3-1

          Gene Ontology Categories:

          Process(es)

          structural molecule activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P107454 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3382399 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P107454 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34534 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 47679 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 6639 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 35625 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 52693 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60386 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 56618 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28568 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 44655 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18752 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 42404 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59050 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3382399 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P107454 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P107454 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3382399 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P107454 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P108840 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P107454 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 8496 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_031958_at - -
          Affymetrix Human Gene 1.0 ST Array 8015174 - -
          Affymetrix Human Gene 1.0 ST Array 8015178 - -
          Affymetrix Human Gene 1.0 ST Array 8019595 - -
          Affymetrix Human Gene 1.0 ST Array 8019596 - -
          Affymetrix Human Gene 1.0 ST Array 8015177 - -
          Affymetrix Human Gene 1.0 ST Array 8015176 - -
          Affymetrix Human Gene 1.0 ST Array 8019594 - -
          Affymetrix Human Gene 1.0 ST Array 8015175 - -
          Affymetrix Human Gene 1.1 ST Array 8015173 - -
          Affymetrix Human Gene 1.1 ST Array 8019593 - -
          Affymetrix HT HG-U133+ PM Array Plate 233537_PM_at - -
          Affymetrix Human Genome U219 Array 11738302_PM_s_at - -
          Affymetrix Human Genome U219 Array 11738302_s_at - -
          Affymetrix Human Genome U219 Array 11738301_at - -
          Affymetrix Human Genome U219 Array 11738301_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR17019656.hg.1 ENST00000391588.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17019656.hg.1 NM_031958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17019656.hg.1 TC17001478.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 NM_031958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 NM_031958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 NM_031958.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010647.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184086.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184086.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184086.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184086.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184086.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184086.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184086.hg.1 TC1700010647.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700184087.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184087.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184087.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184087.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184087.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700184087.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700184087.hg.1 TC1700010647.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000391588.2 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 ENST00000581033.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 NM_031958.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010647.hg.1 NM_031958.1 Refseq
          Human Genome U133 Plus 2.0 Array 233537_PM_at - -
          Human Genome U133 Plus 2.0 Array 233537_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233537_at2 - -
          Human Genome U133 Set 233537_PM_at - -
          Human Genome U133 Set 233537_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008305 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008305 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 133475 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1652439 AJ406931.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1652439 BC113077.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1652439 BC113078.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1652439 NM_031958.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1655251 AJ406931.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1652439 AJ406931.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1652439 BC113077.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1652439 BC113078.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1652439 NM_031958.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1652439 AJ406931.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1652439 BC113077.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1652439 BC113078.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1652439 NM_031958.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1652439 AJ406931.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1652439 BC113077.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1652439 BC113078.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1652439 NM_031958.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_13994361-S AJ406931.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_13994361-S NM_031958.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_23562-S - -
          Illumina human-6 v1.0 expression beadchip 0004670465 - -
          Illumina humanRef-8 v1.0 expression beadchip 0004670465 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1652439 AJ406931.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1652439 BC113077.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1652439 BC113078.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1652439 NM_031958.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 6940541 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_23562 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652439 AJ406931.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652439 BC113077.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652439 BC113078.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1652439 NM_031958.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1652439 AJ406931.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1652439 BC113077.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1652439 BC113078.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1652439 NM_031958.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1655251 NM_031958.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1652439 AJ406931.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1652439 BC113077.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1652439 BC113078.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1652439 NM_031958.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655251 AJ406931.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1655251 NM_031958.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline