Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00541038_m1
          See other SHMT1 GE Assays ›
          Gene Symbol:
          SHMT1
          Gene Name:
          serine hydroxymethyltransferase 1
          Gene Aliases:
          CSHMT, SHMT
          Chromosome Location:
          Chr.17: 18327860 - 18363563 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00541038_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SHMT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001281786.1 NP_001268715.1 1 - 2 197 58
          NM_004169.4 NP_004160.3 1 - 2 197 58
          NM_148918.2 NP_683718.1 1 - 2 197 58
          XM_011523992.2 XP_011522294.1 1 - 2 165 58
          GenBank mRNA AK223552.1 - 1 - 2 128 58
          AK298415.1 - 1 - 2 129 58
          AK302834.1 - 1 - 2 135 58
          BC007979.2 - 1 - 2 43 3687873 58
          BC022874.1 - 1 - 2 47 4108106 58
          BC038598.1 - 1 - 2 54 4523709 58
          Y14485.1 - 1 - 2 163 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700009974.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001201.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK298415.1,AK302834.1,BC007979.2,BC022874.1,BC038598.1,NM_001281786.1,NM_004169.4,NM_148918.2,XM_011523992.2,Y14485.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SHMT1

          Panther Classification:

          Molecular Function -

          methyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          dTMP biosynthetic process
          glycine metabolic process
          L-serine metabolic process
          L-serine catabolic process
          purine nucleobase biosynthetic process
          glycine biosynthetic process from serine
          tetrahydrofolate interconversion
          carnitine biosynthetic process
          tetrahydrofolate metabolic process
          folic acid metabolic process
          protein tetramerization
          protein homotetramerization
          cellular response to tetrahydrofolate
          glycine hydroxymethyltransferase activity
          protein binding
          L-allo-threonine aldolase activity
          amino acid binding
          pyridoxal phosphate binding
          protein homodimerization activity
          serine binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000316694.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000352886.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000354098.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395682.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000395684.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000579558.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000580002.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582352.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000582653.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000583780.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000617912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 ENST00000618902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_001281786.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_004169.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NM_148918.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009974.hg.1 NR_106836.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100013937.hg.1 ENST00000419974.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100013937.hg.1 ENST00000419974.2 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 122104 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gt9tc:80/100.66.77.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0