Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00541513_s1
          See other PIP5K1P1 GE Assays ›
          Gene Symbol:
          PIP5K1P1
          Gene Name:
          phosphatidylinositol-4-phosphate 5-kinase type 1 pseudogene 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 7986102 - 7990344 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00541513_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIP5K1P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027712.1 1 53 85
          GenBank mRNA BC028580.2 - 1 52 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600006877.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000063.hg.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 530642 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 451658 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 88922 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3884610 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P389426 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 39570 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 21415 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3884610 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3884610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725735 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782347 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725744 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538357 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538360 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725741 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538350 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725749 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725750 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725735 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24725742 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782341 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538359 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04538353 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15782347 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P389426 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P125393 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P389426 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538432 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538398 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538350 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782356 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782384 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782376 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782383 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538391 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538357 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782372 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538352 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538429 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538370 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782354 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538393 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538418 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538372 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538416 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538363 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538366 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782377 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538384 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538426 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538377 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538360 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538365 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538346 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538411 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538355 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15782336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538367 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04538387 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8116828 - -
          Affymetrix Human Gene 1.1 ST Array 8116827 - -
          Affymetrix Human Transcriptome Array 2.0 PSR06000615.hg.1 TC06000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000615.hg.1 NR_027712.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06000616.hg.1 TC06000063.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06000616.hg.1 ENST00000438779.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06000616.hg.1 NR_027712.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006877.hg.1 ENST00000460661.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600006877.hg.1 NR_027712.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006877.hg.1 NR_027712.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600006877.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006877.hg.1 ENST00000460661.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600006877.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006877.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay TC0600006877.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600147101.hg.1 ENST00000460661.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147101.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600147101.hg.1 ENST00000460661.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600147101.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600147101.hg.1 TC0600006877.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600147100.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600147100.hg.1 NR_027712.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600147100.hg.1 TC0600006877.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030959 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030960 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030960 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030959 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 205196 - -
          ABI Human Genome Survey Microarray Version 2.0 211262 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mqb7j:80/100.66.77.84:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0