| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003499.hg.1 |
ENST00000376705.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003499.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003499.hg.1 |
TC13000337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300007761.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003500.hg.1 |
ENST00000376705.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003500.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003500.hg.1 |
TC13000337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003498.hg.1 |
ENST00000376705.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003498.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003498.hg.1 |
TC13000337.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003501.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13003501.hg.1 |
TC13000337.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1300152285.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152285.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152285.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152285.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152285.hg.1 |
TC1300007761.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300007761.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152287.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152287.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152287.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152287.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152287.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152287.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152287.hg.1 |
TC1300007761.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300152288.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152288.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152288.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152288.hg.1 |
ENST00000376705.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152288.hg.1 |
ENST00000620595.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300152288.hg.1 |
NM_153456.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300152288.hg.1 |
TC1300007761.hg.1 |
Clariom D TC |