Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00543397_m1
          See other LINC00324 GE Assays ›
          Gene Symbol:
          LINC00324
          Gene Name:
          long intergenic non-protein coding RNA 324
          Gene Aliases:
          C17orf44, NCRNA00324
          Chromosome Location:
          Chr.17: 8220630 - 8224043 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00543397_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00324
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026951.1 2 - 3 1215 67
          GenBank mRNA AK092109.1 - 2 - 3 1215 67
          BC063002.1 - 1 - 2 773 67
          BC089396.1 - 1 - 2 744 67
          BC113564.1 - 2 - 3 1056 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700009680.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001110.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 61698 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 60807 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 29618 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 8030 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P362191 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4868 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4869 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 43394 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 11503 - -
          Agilent Human Genome CGH Microarray 44B A_14_P109268 - -
          Agilent Human Genome CGH Microarray 44A A_14_P109268 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 22443 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P362191 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P362191 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216981 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216964 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767934 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216965 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216977 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216963 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216969 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216965 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216981 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216972 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216964 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767919 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216980 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216974 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767934 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30767935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10216977 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P109268 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P109268 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P109268 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P390650 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P362191 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 139363 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 42275 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P109268 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P34048654 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216942 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216968 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216979 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216969 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216982 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216975 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216944 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216958 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216972 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216983 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216963 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216959 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216940 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216965 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216960 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216977 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16941455 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216981 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10216938 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P109268 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8012418 - -
          Affymetrix Human Gene 1.0 ST Array 8012420 - -
          Affymetrix Human Gene 1.0 ST Array 8012421 - -
          Affymetrix Human Gene 1.0 ST Array 8012417 - -
          Affymetrix Human Gene 1.0 ST Array 8012422 - -
          Affymetrix Human Gene 1.0 ST Array 8012419 - -
          Affymetrix Human Gene 1.1 ST Array 8012416 - -
          Affymetrix HT HG-U133+ PM Array Plate 230012_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR17014950.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17014950.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17014950.hg.1 TC17001110.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17014948.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17014948.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17014948.hg.1 TC17001110.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17014949.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR17014949.hg.1 NR_026951.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17014949.hg.1 TC17001110.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700175429.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175429.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175429.hg.1 TC1700009680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700175429.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175429.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175430.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175430.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175430.hg.1 TC1700009680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700175430.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175430.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay TC1700009680.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175427.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175427.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175427.hg.1 TC1700009680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700175427.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175427.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175428.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175428.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175428.hg.1 TC1700009680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700175428.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175428.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175426.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175426.hg.1 NR_026951.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700175426.hg.1 TC1700009680.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700175426.hg.1 ENST00000315707.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1700175426.hg.1 NR_026951.1 Refseq
          Human Genome U133 Plus 2.0 Array 230012_at - -
          Human Genome U133 Plus 2.0 Array 230012_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 230012_at1 - -
          Human Genome U95 Set 73657_PM_at - -
          Human Genome U95 Set 73657_at - -
          Human Genome U133 Set 230012_at - -
          Human Genome U133 Set 230012_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007876 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A007876 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 225269 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681252 AK092109.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681252 BC063002.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681252 BC089396.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1681252 NR_026951.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1681252 AK092109.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1681252 BC063002.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1681252 BC089396.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1681252 NR_026951.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1681252 AK092109.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1681252 BC063002.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1681252 BC089396.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1681252 NR_026951.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681252 AK092109.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681252 BC063002.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681252 BC089396.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1681252 NR_026951.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1681252 AK092109.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1681252 BC063002.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1681252 BC089396.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1681252 NR_026951.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline