Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00543406_m1
          See other KRT18P55 GE Assays ›
          Gene Symbol:
          KRT18P55
          Gene Name:
          keratin 18 pseudogene 55
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 28275986 - 28307382 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00543406_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KRT18P55
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028334.1 2 - 3 521 109
          GenBank mRNA AK097823.1 - 2 - 3 522 109
          BC112026.1 - 2 - 3 142 8327494 109
          BC112028.1 - 2 - 3 130 8327496 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700010186.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17001293.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK097823.1,BC112026.1,BC112028.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P373708 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 2931 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 24574 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P20616170 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P20616145 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P20616207 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P373708 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P373708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835625 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835629 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271484 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835648 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835654 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835623 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835647 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835641 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835636 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835627 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835643 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835642 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835625 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835654 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835629 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835643 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835636 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271489 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835655 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835645 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271499 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835627 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271493 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835656 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835647 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835641 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835623 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835642 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271492 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835648 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835639 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835653 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271484 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P373708 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P133054 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P106924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8013577 - -
          Affymetrix Human Gene 1.0 ST Array 8013579 - -
          Affymetrix Human Gene 1.0 ST Array 8013578 - -
          Affymetrix Human Gene 1.0 ST Array 8013576 - -
          Affymetrix Human Gene 1.0 ST Array 8013575 - -
          Affymetrix Human Gene 1.0 ST Array 8013580 - -
          Affymetrix Human Gene 1.1 ST Array 8013574 - -
          Affymetrix Human Genome U219 Array 11735557_x_at - -
          Affymetrix Human Genome U219 Array 11735557_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR17017064.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17017064.hg.1 TC17001293.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17017068.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17017068.hg.1 TC17001293.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR17017065.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17017065.hg.1 TC17001293.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000385109.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179613.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179613.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179613.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179613.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179613.hg.1 TC1700010186.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700179608.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179608.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179608.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179608.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179608.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179608.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179608.hg.1 TC1700010186.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700179614.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179614.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179614.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179614.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179614.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179614.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179614.hg.1 TC1700010186.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179607.hg.1 TC1700010186.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700179610.hg.1 TC1700010186.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000385109.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000577198.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000578578.5 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 ENST00000581956.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Affymetrix Human Clariom D Assay TC1700010186.hg.1 NR_028334.1 Refseq
          Human Genome U95 Set 72846_at - -
          Human Genome U95 Set 72846_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008098 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A008098 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 220065 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1722537 NR_028334.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1722537 AK097823.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2041222 NR_028334.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2041222 AK097823.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1722537 NR_028334.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1722537 AK097823.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1722537 NR_028334.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1722537 AK097823.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2041222 NR_028334.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2041222 AK097823.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1722537 NR_028334.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1722537 AK097823.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2041222 NR_028334.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2041222 AK097823.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2041222 NR_028334.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2041222 AK097823.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1722537 NR_028334.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1722537 AK097823.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2041222 NR_028334.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2041222 AK097823.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline