Search
Search

HIST1H3A| Gene Symbol: HIST1H3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003529.2 | NP_003520.1 | 1 | 426 | 73 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812585 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04624358 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812579 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24812578 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04624361 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P111037 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P111037 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 45193 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P120482 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P120482 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 31064 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P01445138 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P111037 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P111037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812582 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812586 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624365 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04624366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24812585 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P111037 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P120482 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04624367 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04624367 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04624366 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P120482 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P120482 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P111037 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 3612 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 57086 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 62629 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624374 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624412 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624421 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624378 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793585 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624419 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624408 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624414 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624364 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624417 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624403 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624396 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624352 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624344 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624394 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624391 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624386 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624406 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624321 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624346 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624363 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624350 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624357 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624388 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624349 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624329 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624410 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624397 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624367 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624371 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04624359 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_003529_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8117331 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8117332 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8117330 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 208575_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11715121_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11715121_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11759077_at | - | - |
| Affymetrix Human Genome U219 Array | 11759077_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208506_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208496_x_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208506_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208575_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208577_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214472_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208577_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214472_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214616_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208576_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208575_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208576_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214616_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208496_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 208575_at | - | - |
| Human Genome U133A 2.0 Array | 214616_at | - | - |
| Human Genome U133A 2.0 Array | 208576_s_at | - | - |
| Human Genome U133A 2.0 Array | 214616_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214472_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208577_at | - | - |
| Human Genome U133A 2.0 Array | 214472_at | - | - |
| Human Genome U133A 2.0 Array | 208575_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208496_x_at | - | - |
| Human Genome U133A 2.0 Array | 208496_PM_x_at | - | - |
| Human Genome U133A 2.0 Array | 208506_at | - | - |
| Human Genome U133A 2.0 Array | 208506_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208576_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 208577_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003529_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw588432 | - | - |
| Human Genome U95 Set | 35598_PM_at | - | - |
| Human Genome U95 Set | 35598_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208575_at11 | - | - |
| Human Genome U133 Set | 214472_PM_at | - | - |
| Human Genome U133 Set | 214472_at | - | - |
| Human Genome U133 Set | 208577_PM_at | - | - |
| Human Genome U133 Set | 208575_at | - | - |
| Human Genome U133 Set | 208496_x_at | - | - |
| Human Genome U133 Set | 208506_at | - | - |
| Human Genome U133 Set | 208506_PM_at | - | - |
| Human Genome U133 Set | 208575_PM_at | - | - |
| Human Genome U133 Set | 208576_PM_s_at | - | - |
| Human Genome U133 Set | 208576_s_at | - | - |
| Human Genome U133 Set | 208577_at | - | - |
| Human Genome U133 Set | 214616_at | - | - |
| Human Genome U133 Set | 208496_PM_x_at | - | - |
| Human Genome U133 Set | 214616_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031091 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031091 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 101193 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_19743828-S | BC066247.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_19743828-S | BC067490.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_19743828-S | BC067491.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_19743828-S | BC069303.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_19743828-S | BC066246.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_28422-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006040088 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006040088 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5860360 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_28422 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1776845 | BC066246.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1776845 | BC066247.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1776845 | BC067490.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1776845 | BC067491.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1776845 | BC069303.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1776845 | BC066246.1 | human_transcriptome |