| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8025131 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8033354 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8033357 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8033355 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8025128 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8033360 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8025130 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8025129 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8025126 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8033359 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8033356 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8033352 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
1552456_PM_a_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000358469.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000358469.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000358469.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001873.hg.1 |
ENST00000381393.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001873.hg.1 |
ENST00000412046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001873.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001873.hg.1 |
TC19000123.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001874.hg.1 |
ENST00000381393.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001874.hg.1 |
ENST00000412046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001874.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001874.hg.1 |
TC19000123.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001875.hg.1 |
ENST00000381393.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001875.hg.1 |
ENST00000412046.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001875.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001875.hg.1 |
TC19000123.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001877.hg.1 |
ENST00000381393.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001877.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19001877.hg.1 |
TC19000123.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148015.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148015.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148015.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148015.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148015.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148015.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148015.hg.1 |
TC1900006834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000381394.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
ENST00000619943.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009455.hg.1 |
NM_001164419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148016.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148016.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148016.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148016.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148016.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148016.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148016.hg.1 |
TC1900006834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900148018.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148018.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148018.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148018.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148018.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148018.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148018.hg.1 |
TC1900006834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006834.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148014.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148014.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148014.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148014.hg.1 |
ENST00000381393.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148014.hg.1 |
ENST00000616171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148014.hg.1 |
NM_144614.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148014.hg.1 |
TC1900006834.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000329753.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
ENST00000618571.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006833.hg.1 |
NM_001136507.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000358469.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000358469.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000358469.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009454.hg.1 |
ENST00000612322.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000333843.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
ENST00000611585.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009456.hg.1 |
NM_001164425.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
1552456_PM_a_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
1552456_a_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
1552456_a_at9 |
- |
- |