| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00928487 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15204585 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002571 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002564 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00928479 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002566 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002562 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002578 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002565 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002592 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002583 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00928490 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002572 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002576 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002595 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P00928471 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002585 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P21002563 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 4x180K |
170134 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 8x60K |
44336 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
77876 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P303238 |
- |
- |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P303238 |
- |
- |
| Agilent Whole Human Genome Oligo Microarray 4x44K |
31768 |
- |
- |
| Agilent Human Genome CGH Microarray 44B |
A_14_P127091 |
- |
- |
| Agilent Human Genome CGH Microarray 44A |
A_14_P127091 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
14721 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_14_P127091 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P303238 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_23_P303238 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928479 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002565 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928493 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002566 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002583 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002574 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15204585 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002570 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928492 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002571 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002585 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002578 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928477 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002572 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928496 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002562 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002576 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002563 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002568 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002564 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928490 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002566 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002583 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928479 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002570 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002578 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002564 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002585 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15204585 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002563 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002565 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928490 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002574 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002568 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928477 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002562 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928492 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928496 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002576 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002571 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P21002572 |
- |
- |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P00928493 |
- |
- |
| Whole Human Genome Oligo Microarray G4112A |
A_23_P303238 |
- |
- |
| Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P127091 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P127091 |
- |
- |
| Agilent Human Genome CGH Microarray 4x44K |
A_14_P127091 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P303238 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AF085858.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AF322907.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AF380302.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AF395588.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AK025916.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AK026066.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AL832983.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
AY049721.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
NM_022455.3 |
refseq_mrna_human_NM |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
122538 |
NM_172349.1 |
refseq_mrna_human_NM |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
32094 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_14_P127091 |
- |
- |
| Agilent Human 1B Microarray G4111A |
A_32_P332407 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928483 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928492 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928496 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928475 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928498 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928473 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928501 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928479 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928485 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928488 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15204585 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928477 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928476 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928495 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928486 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928481 |
- |
- |
| Agilent Human Whole Genome ChIP-on-Chip |
A_17_P00928490 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_14_P127091 |
- |
- |