| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002108.hg.1 |
ENST00000333598.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002108.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002108.hg.1 |
TC19000134.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002107.hg.1 |
ENST00000333598.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002107.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002107.hg.1 |
TC19000134.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199277.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199277.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199277.hg.1 |
TC1900011657.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199277.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199277.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199274.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199274.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199274.hg.1 |
TC1900011657.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199274.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199274.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
ENST00000598851.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011657.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199276.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199276.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199276.hg.1 |
NM_174918.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199276.hg.1 |
TC1900011657.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199276.hg.1 |
ENST00000333598.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199276.hg.1 |
ENST00000597445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199276.hg.1 |
NM_174918.2 |
Refseq |