| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000588137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000588137.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
ENST00000602211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000263372.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000588137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000588137.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
ENST00000602211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008011.hg.1 |
NM_004823.1 |
Refseq |