Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00601656_s1
          See other HIST1H2AA GE Assays ›
          Gene Symbol:
          HIST1H2AA
          Gene Name:
          histone cluster 1, H2aa
          Gene Aliases:
          H2AA, H2AFR, TH2A, bA317E16.2
          Chromosome Location:
          Chr.6: 25726063 - 25726562 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00601656_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HIST1H2AA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_170745.3 NP_734466.1 1 190 160
          GenBank mRNA BC062211.1 - 1 194 6614487 160

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011109.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001335.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HIST1H2AA

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin silencing
          DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04622999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811260 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811289 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623007 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811302 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811283 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623031 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623027 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15793466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811301 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15793463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811294 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24811287 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15793464 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04623042 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 74832 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 24266 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P355439 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P355439 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 7266 - -
          Agilent Human Genome CGH Microarray 44B A_14_P113991 - -
          Agilent Human Genome CGH Microarray 44A A_14_P113991 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49697 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P17484724 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P355439 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P355439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811289 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623031 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811302 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623013 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811273 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811289 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623031 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04623040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24811286 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P355439 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P113991 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623020 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623033 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623040 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623019 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623028 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623039 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623030 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623024 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623015 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623031 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623047 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623049 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623044 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623037 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623017 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623042 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04623026 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04623045 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04623043 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04623046 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04623044 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P113991 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P113991 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P355439 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 9406 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 98000 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 48314 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P113991 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623024 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623022 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623004 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623030 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793464 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793463 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793466 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623042 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623018 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623049 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793468 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623037 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623002 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623020 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623026 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623014 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623008 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04623044 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P113991 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_170745_at - -
          Affymetrix Human Gene 1.0 ST Array 8124334 - -
          Affymetrix Human Gene 1.0 ST Array 8124333 - -
          Affymetrix Human Gene 1.0 ST Array 8124332 - -
          Affymetrix Human Gene 1.1 ST Array 8124331 - -
          Affymetrix Human Genome U219 Array 11740813_PM_at - -
          Affymetrix Human Genome U219 Array 11740813_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06016021.hg.1 TC06001335.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016021.hg.1 ENST00000297012.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016021.hg.1 NM_170745.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016020.hg.1 TC06001335.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016020.hg.1 ENST00000297012.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016020.hg.1 NM_170745.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175132.hg.1 NM_170745.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175132.hg.1 NM_170745.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175132.hg.1 TC0600011109.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175133.hg.1 NM_170745.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175133.hg.1 NM_170745.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175133.hg.1 TC0600011109.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029512 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029512 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 117083 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1694166 NM_170745.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_29171724-S NM_170745.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_29171724-S BC062211.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_15137-S - -
          Illumina human-6 v1.0 expression beadchip 0003840497 - -
          Illumina humanRef-8 v1.0 expression beadchip 0003840497 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 870092 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_15137 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1694166 BC062211.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1694166 BC062211.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1694166 NM_170745.3 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1694166 BC062211.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline