Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00602051_mH
          See other FSCN1 GE Assays ›
          Gene Symbol:
          FSCN1
          Gene Name:
          fascin actin-bundling protein 1
          Gene Aliases:
          FAN1, HSN, SNL, p55
          Chromosome Location:
          Chr.7: 5592805 - 5606656 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00602051_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FSCN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003088.3 NP_003079.1 1 - 2 969 126
          GenBank mRNA AK095254.1 - 2 - 3 903 126
          AK316607.1 - 1 - 2 964 126
          BC000521.2 - 1 - 2 860 2961047 126
          BC006304.2 - 1 - 2 951 4100996 126
          BC007539.1 - 1 - 2 860 126
          BC007643.1 - 1 - 2 593 126
          BC007948.2 - 1 - 2 948 4299524 126
          BC007988.2 - 1 - 2 860 2961047 126
          BT006636.1 - 1 - 2 837 126
          U03057.1 - 1 - 2 948 126
          U09873.1 - 1 - 2 882 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700006629.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000055.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens 1700 Inventoried MicroArray Quality Control (MAQC) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FSCN1

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell junction assembly
          cell proliferation
          anatomical structure morphogenesis
          positive regulation of lamellipodium assembly
          cell migration
          microspike assembly
          actin cytoskeleton organization
          regulation of microvillus assembly
          regulation of actin cytoskeleton organization
          establishment of apical/basal cell polarity
          cell motility
          actin filament bundle assembly
          positive regulation of filopodium assembly
          positive regulation of podosome assembly
          positive regulation of extracellular matrix disassembly
          cell-cell adhesion
          actin binding
          protein binding
          drug binding
          protein binding, bridging
          poly(A) RNA binding
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07000894.hg.1 TC07000055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07000894.hg.1 ENST00000340250.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000894.hg.1 ENST00000382361.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000894.hg.1 ENST00000535097.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000894.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700145591.hg.1 TC0700006629.hg.1 Clariom D TC
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 150201 - -
          ABI Human Genome Survey Microarray Version 2.0 169875 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline