Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00602355_g1
          See other PPY2P GE Assays ›
          Gene Symbol:
          PPY2P
          Gene Name:
          pancreatic polypeptide 2, pseudogene
          Gene Aliases:
          PPY2
          Chromosome Location:
          Chr.17: 28247444 - 28248289 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00602355_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PPY2P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002181.1 1 - 2 511 105
          GenBank mRNA AF085945.1 - 1 - 2 406 105
          AF222903.1 - 1 - 2 511 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1700007357.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC17000318.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16965214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30835308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10271304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30835307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30835304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10271296 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10271300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30835309 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10271308 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P10271298 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P164253 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P164253 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 30291 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 51393 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P164253 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P164253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835322 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271316 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271296 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835296 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835322 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271316 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835283 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271298 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835307 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835323 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271296 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271281 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835311 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965202 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965205 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965208 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271287 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965214 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271290 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835296 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271312 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16965207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30835277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10271283 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P164253 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P164253 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271304 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271339 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271351 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271300 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271328 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16965234 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271294 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271280 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271327 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271296 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271290 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271345 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271282 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271292 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271286 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271336 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271279 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271343 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271307 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271332 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271288 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16965226 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271331 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271341 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271299 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271319 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16965229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271325 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271347 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10271311 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002181_at - -
          Affymetrix Human Gene 1.0 ST Array 8005838 - -
          Affymetrix Human Gene 1.0 ST Array 8005837 - -
          Affymetrix Human Gene 1.1 ST Array 8005836 - -
          Affymetrix HT HG-U133+ PM Array Plate 220654_PM_at - -
          Affymetrix Human Genome U219 Array 11744806_at - -
          Affymetrix Human Genome U219 Array 11744806_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 NR_002181.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 NR_002181.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 NR_002181.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1700007357.hg.1 NR_002181.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17003828.hg.1 NR_002181.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR17003828.hg.1 TC17000318.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1700152269.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700152269.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700152269.hg.1 NR_002181.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700152269.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700152269.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1700152269.hg.1 NR_002181.1 Refseq
          Affymetrix Human Clariom D Assay PSR1700152269.hg.1 TC1700007357.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000582808.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 ENST00000583761.1 Ensembl
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 NR_002181.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 NR_002181.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 NR_002181.1 Refseq
          Affymetrix Human Clariom D Assay TC1700007357.hg.1 NR_002181.1 Refseq
          Human Genome U133 Plus 2.0 Array 220654_PM_at - -
          Human Genome U133 Plus 2.0 Array 220654_at - -
          Human Genome U133A 2.0 Array 220654_at - -
          Human Genome U133A 2.0 Array 220654_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002181_at - -
          AffymetrixPathwork Diagnostics Pathchip pw521845 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220654_at9 - -
          Human Genome U95 Set 63453_PM_at - -
          Human Genome U95 Set 63453_at - -
          Human Genome U133 Set 220654_at - -
          Human Genome U133 Set 220654_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009297 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A009297 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 155974 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2136926 NR_002181.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2136926 AF085945.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2136926 AF222903.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1728413 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2136926 NR_002181.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2136926 AF085945.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2136926 AF222903.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2136926 NR_002181.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2136926 AF085945.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2136926 AF222903.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2136926 NR_002181.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2136926 AF085945.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2136926 AF222903.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1728413 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 5960358 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_14558 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2136926 NR_002181.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2136926 AF085945.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2136926 AF222903.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2136926 NR_002181.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2136926 AF085945.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2136926 AF222903.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2136926 NR_002181.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2136926 AF085945.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2136926 AF222903.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1728413 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2136926 NR_002181.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2136926 AF085945.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2136926 AF222903.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline