Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00602694_mH
          See other SRD5A1 GE Assays ›
          Gene Symbol:
          SRD5A1
          Gene Name:
          steroid 5 alpha-reductase 1
          Gene Aliases:
          S5AR 1
          Chromosome Location:
          Chr.5: 6633440 - 6674386 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00602694_mH
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SRD5A1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001047.4 NP_001038.1 2 - 3 601 120
          NM_001324323.2 NP_001311252.1 3 - 4 1103 120
          GenBank mRNA AF052126.1 - 2 - 3 591 120
          AF113128.1 - 3 - 4 478 120
          AK297208.1 - 3 - 4 1116 120
          AK313070.1 - 2 - 3 601 120
          BC006373.2 - 2 - 3 572 120
          BC007033.1 - 2 - 3 601 120
          BC008673.2 - 2 - 3 575 120
          BT006834.1 - 2 - 3 464 120
          M32313.1 - 2 - 3 494 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500013149.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05000044.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Human Androgen Panel Inventoried MicroArray Quality Control (MAQC) Human_Androgen Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SRD5A1

          Gene Ontology Categories:

          Function(s) Process(es)

          urogenital system development
          liver development
          lipid metabolic process
          steroid biosynthetic process
          androgen biosynthetic process
          androgen catabolic process
          sex determination
          sex differentiation
          steroid metabolic process
          androgen metabolic process
          male gonad development
          cellular response to starvation
          response to xenobiotic stimulus
          response to muscle activity
          diterpenoid metabolic process
          spinal cord development
          hippocampus development
          thalamus development
          hypothalamus development
          pituitary gland development
          cerebral cortex development
          cell differentiation
          male genitalia development
          female genitalia development
          response to follicle-stimulating hormone
          response to estradiol
          cellular response to insulin stimulus
          response to testosterone
          serotonin metabolic process
          progesterone metabolic process
          response to estrogen
          bone development
          response to growth hormone
          response to fungicide
          cellular response to cAMP
          cellular response to growth factor stimulus
          cellular response to estradiol stimulus
          cellular response to testosterone stimulus
          cellular response to dexamethasone stimulus
          cellular response to epinephrine stimulus
          cellular response to serotonin
          3-oxo-5-alpha-steroid 4-dehydrogenase activity
          protein binding
          electron carrier activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-CH group of donors
          amide binding
          cholestenone 5-alpha-reductase activity
          NADPH binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05000528.hg.1 ENST00000274192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000528.hg.1 ENST00000537411.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000528.hg.1 ENST00000538824.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000528.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000528.hg.1 TC05000044.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05000529.hg.1 ENST00000274192.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000529.hg.1 ENST00000537411.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000529.hg.1 ENST00000538824.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05000529.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05000529.hg.1 TC05000044.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192671.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192671.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192671.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192671.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192671.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192671.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192671.hg.1 TC0500013149.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000504286.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay TC0500013149.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192672.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192672.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192672.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192672.hg.1 ENST00000510531.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192672.hg.1 ENST00000513117.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500192672.hg.1 NM_001047.3 Refseq
          Affymetrix Human Clariom D Assay PSR0500192672.hg.1 TC0500013149.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline