Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00602749_m1
          See other ZIC1 GE Assays ›
          Gene Symbol:
          ZIC1
          Gene Name:
          Zic family member 1
          Gene Aliases:
          CRS6, ZIC, ZNF201
          Chromosome Location:
          Chr.3: 147409394 - 147416719 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00602749_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ZIC1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003412.3 NP_003403.2 1 - 2 1700 149
          GenBank mRNA BC104848.1 - 1 - 2 1054 8143851 149
          BC104850.1 - 1 - 2 1055 8143853 149
          BF920299.1 - 2 - 3 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300013886.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03000791.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Human Hedgehog Pathway Inventoried Human_Hedgehog Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ZIC1

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          pattern specification process
          brain development
          adult walking behavior
          regulation of smoothened signaling pathway
          spinal cord development
          cell differentiation
          positive regulation of protein import into nucleus
          inner ear morphogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03014269.hg.1 TC03000791.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03014269.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03014269.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03014266.hg.1 TC03000791.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03014266.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03014266.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03014270.hg.1 TC03000791.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03014270.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03014270.hg.1 NM_003412.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03014267.hg.1 TC03000791.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03014267.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03014267.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205588.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205588.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205588.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205588.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205588.hg.1 TC0300013886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay TC0300013886.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205587.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205587.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205587.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205587.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205587.hg.1 TC0300013886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300205589.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205589.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205589.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205589.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205589.hg.1 TC0300013886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300205590.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205590.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205590.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205590.hg.1 ENST00000282928.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205590.hg.1 ENST00000488404.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0300205590.hg.1 NM_003412.3 Refseq
          Affymetrix Human Clariom D Assay PSR0300205590.hg.1 TC0300013886.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline