Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00603461_s1
          See other TAS2R41 GE Assays ›
          Gene Symbol:
          TAS2R41
          Gene Name:
          taste 2 receptor member 41
          Gene Aliases:
          T2R41, T2R59
          Chromosome Location:
          Chr.7: 143477873 - 143478796 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00603461_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TAS2R41
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176883.2 NP_795364.2 1 758 96
          GenBank mRNA BC101157.3 - 1 758 40015710 96
          BC101158.2 - 1 758 40015713 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700009501.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000938.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TAS2R41

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840777 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840739 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840746 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840743 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840768 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840757 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840734 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101078 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26101106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840774 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05840778 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 129025 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 12444 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P337867 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P337867 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34494 - -
          Agilent Human Genome CGH Microarray 44B A_14_P119806 - -
          Agilent Human Genome CGH Microarray 44A A_14_P119806 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 2501 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P32162837 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P337867 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P337867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840724 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840728 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840739 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840734 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840706 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840726 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840730 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840719 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840715 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101044 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840705 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840737 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P26101054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840725 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840708 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05840724 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P337867 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P119806 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840718 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840731 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840734 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840732 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840717 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840719 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840739 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840714 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840728 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840730 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840737 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840726 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840708 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840722 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840715 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840742 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840724 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P05840711 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P119806 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P119806 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P337867 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 2016 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 98319 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P119806 - -
          Agilent Human 1B Microarray G4111A A_32_P343482 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840731 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840771 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840759 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840773 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840750 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840734 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840726 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840776 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840714 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840708 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840743 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840779 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840777 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840737 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840753 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840717 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840739 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840756 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840732 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840769 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840728 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840764 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840730 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840760 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840745 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840748 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840706 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840766 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840711 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840722 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840765 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840704 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05840724 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P119806 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_176883_at - -
          Affymetrix Human Gene 1.0 ST Array 8136935 - -
          Affymetrix Human Gene 1.1 ST Array 8136934 - -
          Affymetrix HT HG-U133+ PM Array Plate 1553558_PM_at - -
          Affymetrix Human Genome U219 Array 11735985_at - -
          Affymetrix Human Genome U219 Array 11735985_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0700009501.hg.1 ENST00000408916.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700009501.hg.1 NM_176883.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07014505.hg.1 TC07000938.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07014505.hg.1 ENST00000408916.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07014505.hg.1 NM_176883.2 Refseq
          Affymetrix Human Clariom D Assay TC0700009501.hg.1 ENST00000408916.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700009501.hg.1 NM_176883.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700167534.hg.1 ENST00000408916.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167534.hg.1 NM_176883.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700167534.hg.1 ENST00000408916.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700167534.hg.1 NM_176883.2 Refseq
          Affymetrix Human Clariom D Assay PSR0700167534.hg.1 TC0700009501.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1553558_PM_at - -
          Human Genome U133 Plus 2.0 Array 1553558_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_176883_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1553558_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035234 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A035234 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1693303 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1693303 - -
          HumanHT-12 V4.0 expression beadchip ILMN_1693303 - -
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1693303 - -
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1693303 - -
          Sentrix Human-6 Expression BeadChip GI_28882027-S - -
          Illumina HumanRef-8 v2 Expression BeadChip GI_27956-S - -
          Illumina human-6 v1.0 expression beadchip 0001240114 - -
          Illumina humanRef-8 v1.0 expression beadchip 0001240114 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1693303 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1693303 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 4010156 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_27956 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1693303 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1693303 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1693303 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline