Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00605586_m1
          See other PAK2 GE Assays ›
          Gene Symbol:
          PAK2
          Gene Name:
          p21 (RAC1) activated kinase 2
          Gene Aliases:
          PAK65, PAKgamma
          Chromosome Location:
          Chr.3: 196739857 - 196832647 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00605586_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PAK2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002577.4 NP_002568.2 7 - 8 1029 143
          XM_011512870.2 XP_011511172.1 7 - 8 752 143
          XM_017006501.1 XP_016861990.1 8 - 9 1216 143
          GenBank mRNA AK291339.1 - 7 - 8 913 143
          U24153.1 - 6 - 7 746 143
          U25975.1 - 6 - 7 614 143

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300010033.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001100.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Homo sapiens Human MAP Kinase Pathways Human PDGF Pathway Human TNFR1 Pathway HUMAN FAS SIGNALING HUMAN GPCR PATHWAY HUMAN GUIDANCE CUES AND GROWTH CONE MOTILITY HUMAN RHO FAMILY GTPASES HUMAN RAS PATHWAY Inventoried Human_PDGF_Pathway Human_Map_Kinase Human_TNFR1_Pathway Human_Guidance_Cues_and_Growth_Cone_Motility Human_GPCR_Pathway Human_Fas_Signaling Human_Ras_Pathway Human_Rho_Family_GTPases Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PAK2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          stimulatory C-type lectin receptor signaling pathway
          protein phosphorylation
          negative regulation of protein kinase activity
          apoptotic process
          signal transduction
          viral process
          phosphorylation
          peptidyl-serine phosphorylation
          signal transduction by protein phosphorylation
          T cell costimulation
          Fc-epsilon receptor signaling pathway
          regulation of growth
          negative regulation of apoptotic process
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          regulation of defense response to virus by virus
          positive regulation of peptidyl-tyrosine phosphorylation
          T cell receptor signaling pathway
          dendritic spine development
          positive regulation of protein tyrosine kinase activity
          cellular response to organic cyclic compound
          cell-cell adhesion
          positive regulation of extrinsic apoptotic signaling pathway
          negative regulation of cysteine-type endopeptidase activity involved in execution phase of apoptosis
          protein kinase activity
          protein serine/threonine kinase activity
          receptor signaling protein serine/threonine kinase activity
          protein binding
          ATP binding
          protein kinase binding
          protein tyrosine kinase activator activity
          identical protein binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR03019019.hg.1 TC03001100.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019019.hg.1 ENST00000327134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019019.hg.1 ENST00000488224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019019.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019011.hg.1 TC03001100.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019011.hg.1 ENST00000327134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019011.hg.1 ENST00000488224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019011.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019012.hg.1 TC03001100.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019012.hg.1 ENST00000327134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019012.hg.1 ENST00000488224.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019012.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019014.hg.1 TC03001100.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019014.hg.1 ENST00000327134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019014.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019018.hg.1 TC03001100.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019018.hg.1 ENST00000327134.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019018.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173081.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173081.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173081.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173081.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173081.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173081.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173081.hg.1 TC0300010033.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay TC0300010033.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173069.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173069.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173069.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173069.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173069.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173069.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173069.hg.1 TC0300010033.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173071.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173071.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173071.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173071.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173071.hg.1 ENST00000481344.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173071.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173071.hg.1 TC0300010033.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173074.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173074.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173074.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173074.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173074.hg.1 TC0300010033.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173079.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173079.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173079.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173079.hg.1 ENST00000327134.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173079.hg.1 ENST00000426668.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173079.hg.1 NM_002577.4 Refseq
          Affymetrix Human Clariom D Assay PSR0300173079.hg.1 TC0300010033.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0