Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00605772_s1
          See other H2BC14 GE Assays ›
          Gene Symbol:
          H2BC14
          Gene Name:
          H2B clustered histone 14
          Gene Aliases:
          H2B/e, H2BFE, HIST1H2BM, dJ160A22.3
          Chromosome Location:
          Chr.6: 27815022 - 27815489 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00605772_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2BC14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003521.3 NP_003512.1 1 71 85
          GenBank mRNA BC066244.1 - 1 49 85
          BC067485.1 - 1 49 85
          BC067486.1 - 1 49 85
          BC067488.1 - 1 49 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007377.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000265.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2BC14

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA binding
          structural constituent of chromatin
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15796474 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P3783 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P3783 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 25741 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19804 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P37558730 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P3783 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P3783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04630051 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P3783 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P3783 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 6186 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 10409 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630030 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630072 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630071 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630077 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15796474 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630085 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630079 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630075 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630087 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04630069 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_003521_at - -
          Affymetrix Human Gene 1.0 ST Array 8117596 - -
          Affymetrix Human Gene 1.0 ST Array 8117597 - -
          Affymetrix Human Gene 1.0 ST Array 8117595 - -
          Affymetrix Human Gene 1.1 ST Array 8117594 - -
          Affymetrix HT HG-U133+ PM Array Plate 208515_PM_at - -
          Affymetrix Human Genome U219 Array 11733740_at - -
          Affymetrix Human Genome U219 Array 11733740_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06002226.hg.1 ENST00000359465.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06002226.hg.1 NM_003521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06002226.hg.1 TC06000265.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0600007377.hg.1 ENST00000621112.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007377.hg.1 NM_003521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007377.hg.1 NM_003521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007377.hg.1 NM_003521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06002230.hg.1 ENST00000359465.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06002230.hg.1 NM_003521.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06002230.hg.1 TC06000265.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0600007377.hg.1 ENST00000621112.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007377.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007377.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007377.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149994.hg.1 ENST00000621112.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600149994.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149994.hg.1 ENST00000621112.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600149994.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149994.hg.1 TC0600007377.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600149996.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149996.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149996.hg.1 TC0600007377.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600149995.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149995.hg.1 NM_003521.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600149995.hg.1 TC0600007377.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 208515_PM_at - -
          Human Genome U133 Plus 2.0 Array 208515_at - -
          Human Genome U133A 2.0 Array 208515_at - -
          Human Genome U133A 2.0 Array 208515_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_003521_at - -
          AffymetrixPathwork Diagnostics Pathchip pw527479 - -
          Human Genome U95 Set 31528_f_at - -
          Human Genome U95 Set 31528_PM_f_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208515_at8 - -
          Human Genome U133 Set 208515_at - -
          Human Genome U133 Set 208515_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031154 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A031154 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 193947 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1736820 BC067486.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1736820 BC067487.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1736820 BC067489.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_15718721-S BC067486.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_15718721-S BC067488.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_15718721-S BC067489.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_15718721-S NM_003521.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_15718721-S BC066244.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_27454-S - -
          Illumina human-6 v1.0 expression beadchip 0005390446 - -
          Illumina humanRef-8 v1.0 expression beadchip 0005390446 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 3990343 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_27454 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1736820 NM_003521.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1736820 BC067486.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1736820 BC067487.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1736820 BC067489.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1736820 NM_003521.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline