Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00605810_s1
          See other HIST1H3B GE Assays ›
          Gene Symbol:
          HIST1H3B
          Gene Name:
          histone cluster 1, H3b
          Gene Aliases:
          H3/l, H3FL
          Chromosome Location:
          Chr.6: 26031589 - 26032060 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00605810_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HIST1H3B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003537.3 NP_003528.1 1 339 149
          GenBank mRNA BC069133.1 - 1 339 7216989 149
          BC096132.1 - 1 339 40002522 149
          BC096133.3 - 1 339 40002523 149
          BC096134.3 - 1 339 40002524 149
          BC099630.3 - 1 342 40002521 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011124.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001342.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Human Interferon Pathway 1700 Inventoried Human_Interferon_Pathway Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HIST1H3B

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin silencing at rDNA
          positive regulation of defense response to virus by host
          nucleosome assembly
          DNA replication-dependent nucleosome assembly
          blood coagulation
          gene silencing by RNA
          telomere organization
          cellular protein metabolic process
          negative regulation of gene expression, epigenetic
          positive regulation of gene expression, epigenetic
          protein heterotetramerization
          regulation of gene silencing
          cell-cell adhesion
          xenophagy
          DNA binding
          protein binding
          histone binding
          protein heterodimerization activity
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624430 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15793591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812667 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 170820 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 32809 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P93258 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P174924 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P93258 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 30039 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 10182 - -
          Agilent Human Genome CGH Microarray 44B A_14_P116959 - -
          Agilent Human Genome CGH Microarray 44A A_14_P116959 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60542 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16761 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P93258 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00317034 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P93258 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812665 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812667 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15793591 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624432 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812665 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15793591 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812667 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P93258 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P174924 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P116959 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624436 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624434 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624430 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624433 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624429 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15793591 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P116959 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P116959 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P174924 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P93258 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 7293 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 6171 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 18955 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 60920 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P116959 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624441 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624377 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793585 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624388 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624410 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624444 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624436 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624374 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624386 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624406 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624439 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624450 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624455 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624384 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624442 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624371 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793591 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624447 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624448 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624458 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624403 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624381 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P116959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_003537_at - -
          Affymetrix Human Gene 1.0 ST Array 8124389 - -
          Affymetrix Human Gene 1.0 ST Array 8124390 - -
          Affymetrix Human Gene 1.1 ST Array 8124388 - -
          Affymetrix HT HG-U133+ PM Array Plate 208576_PM_s_at - -
          Affymetrix Human Genome U219 Array 11732968_at - -
          Affymetrix Human Genome U219 Array 11732968_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06016115.hg.1 TC06001342.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06016115.hg.1 ENST00000244661.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06016115.hg.1 NM_003537.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175277.hg.1 NM_003537.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175277.hg.1 NM_003537.3 Refseq
          Affymetrix Human Clariom D Assay PSR0600175277.hg.1 TC0600011124.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 208496_x_at - -
          Human Genome U133 Plus 2.0 Array 208576_s_at - -
          Human Genome U133 Plus 2.0 Array 208577_PM_at - -
          Human Genome U133 Plus 2.0 Array 208575_PM_at - -
          Human Genome U133 Plus 2.0 Array 214472_at - -
          Human Genome U133 Plus 2.0 Array 214616_PM_at - -
          Human Genome U133 Plus 2.0 Array 208506_at - -
          Human Genome U133 Plus 2.0 Array 208576_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 208496_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 214472_PM_at - -
          Human Genome U133 Plus 2.0 Array 208577_at - -
          Human Genome U133 Plus 2.0 Array 208575_at - -
          Human Genome U133 Plus 2.0 Array 214616_at - -
          Human Genome U133 Plus 2.0 Array 208506_PM_at - -
          Human Genome U133A 2.0 Array 208576_PM_s_at - -
          Human Genome U133A 2.0 Array 214472_at - -
          Human Genome U133A 2.0 Array 214616_PM_at - -
          Human Genome U133A 2.0 Array 208496_PM_x_at - -
          Human Genome U133A 2.0 Array 214616_at - -
          Human Genome U133A 2.0 Array 208577_at - -
          Human Genome U133A 2.0 Array 208576_s_at - -
          Human Genome U133A 2.0 Array 208577_PM_at - -
          Human Genome U133A 2.0 Array 208575_PM_at - -
          Human Genome U133A 2.0 Array 214472_PM_at - -
          Human Genome U133A 2.0 Array 208506_at - -
          Human Genome U133A 2.0 Array 208575_at - -
          Human Genome U133A 2.0 Array 208496_x_at - -
          Human Genome U133A 2.0 Array 208506_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_003537_at - -
          AffymetrixPathwork Diagnostics Pathchip pw595188 - -
          Human Genome U95 Set 35598_PM_at - -
          Human Genome U95 Set 35598_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208576_s_at4 - -
          Human Genome U133 Set 208575_PM_at - -
          Human Genome U133 Set 208576_PM_s_at - -
          Human Genome U133 Set 214616_at - -
          Human Genome U133 Set 208575_at - -
          Human Genome U133 Set 208506_PM_at - -
          Human Genome U133 Set 214472_at - -
          Human Genome U133 Set 208576_s_at - -
          Human Genome U133 Set 208496_PM_x_at - -
          Human Genome U133 Set 208577_PM_at - -
          Human Genome U133 Set 214616_PM_at - -
          Human Genome U133 Set 208506_at - -
          Human Genome U133 Set 214472_PM_at - -
          Human Genome U133 Set 208496_x_at - -
          Human Genome U133 Set 208577_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029524 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029524 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 304298 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2222163 BC096134.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2222163 BC099630.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2222163 NM_003537.3 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2222163 BC069133.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2222163 BC096132.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1752063 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2222163 BC096134.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2222163 BC099630.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2222163 NM_003537.3 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2222163 BC069133.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2222163 BC096132.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2222163 BC096134.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2222163 BC099630.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2222163 NM_003537.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2222163 BC069133.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2222163 BC096132.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2222163 BC096134.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2222163 BC099630.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2222163 NM_003537.3 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2222163 BC069133.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2222163 BC096132.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396498-S BC096134.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396498-S BC099630.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396498-S NM_003537.3 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396498-S BC069133.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_21396498-S BC096132.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_30162-S - -
          Illumina human-6 v1.0 expression beadchip 0004210524 - -
          Illumina humanRef-8 v1.0 expression beadchip 0004210524 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1752063 - -
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1752063 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_30162 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_30162 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2222163 BC096134.3 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2222163 BC099630.3 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2222163 NM_003537.3 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2222163 BC069133.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2222163 BC096132.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2222163 BC096134.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2222163 BC099630.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2222163 NM_003537.3 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2222163 BC069133.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2222163 BC096132.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2222163 BC096134.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2222163 BC099630.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2222163 NM_003537.3 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2222163 BC069133.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2222163 BC096132.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1752063 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2222163 BC096134.3 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2222163 BC099630.3 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2222163 NM_003537.3 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2222163 BC069133.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2222163 BC096132.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0