Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00606684_s1
          See other HIST1H2BB GE Assays ›
          Gene Symbol:
          HIST1H2BB
          Gene Name:
          histone cluster 1, H2bb
          Gene Aliases:
          H2B.1, H2B/f, H2BFF
          Chromosome Location:
          Chr.6: 26043227 - 26043657 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00606684_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HIST1H2BB
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021062.2 NP_066406.1 1 57 140
          GenBank mRNA BC096728.1 - 1 57 40014250 140

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HIST1H2BB

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA binding
          protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24812700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624466 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624467 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04624462 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P111054 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P111054 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 32222 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18467 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01445170 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P111054 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P111054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624466 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624462 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624467 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812700 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624470 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24812707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624462 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624463 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624467 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04624466 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P111054 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624472 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624464 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624475 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624467 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624462 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624469 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624476 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624478 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624466 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P04624473 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624468 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624469 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624470 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624478 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P04624467 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P111054 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 8473 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624442 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624439 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624466 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624447 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624502 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624487 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624492 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624453 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624470 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624448 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624494 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624476 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624478 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624467 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624490 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624444 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624441 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624464 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624482 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624485 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624462 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624450 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624475 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624497 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04624443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15793600 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_021062_at - -
          Affymetrix Human Gene 1.0 ST Array 8124395 - -
          Affymetrix Human Gene 1.0 ST Array 8124396 - -
          Affymetrix Human Gene 1.1 ST Array 8124394 - -
          Affymetrix HT HG-U133+ PM Array Plate 208547_PM_at - -
          Affymetrix Human Genome U219 Array 11738961_at - -
          Affymetrix Human Genome U219 Array 11738962_x_at - -
          Affymetrix Human Genome U219 Array 11738962_PM_x_at - -
          Affymetrix Human Genome U219 Array 11738961_PM_at - -
          Human Genome U133 Plus 2.0 Array 208547_PM_at - -
          Human Genome U133 Plus 2.0 Array 208547_at - -
          Human Genome U133A 2.0 Array 208547_PM_at - -
          Human Genome U133A 2.0 Array 208547_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_021062_at - -
          AffymetrixPathwork Diagnostics Pathchip pw559019 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 208547_at1 - -
          Human Genome U133 Set 208547_at - -
          Human Genome U133 Set 208547_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029527 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029527 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 189299 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1750180 NM_021062.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_19924303-S NM_021062.2 human_transcriptome
          Sentrix Human-6 Expression BeadChip GI_19924303-S BC096728.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_30056-S - -
          Illumina human-6 v1.0 expression beadchip 0002940021 - -
          Illumina humanRef-8 v1.0 expression beadchip 0002940021 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 1230278 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_30056 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750180 BC096728.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1750180 BC096728.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1750180 NM_021062.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1750180 BC096728.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline