Search
Search

KRTAP9-3| Gene Symbol: KRTAP9-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031962.2 | NP_114168.1 | 1 | 670 | 151 | |
| GenBank mRNA | AJ406947.1 | - | 1 | 670 | 151 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893520 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981659 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16981651 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321713 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30893501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321712 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321708 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10321707 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 174235 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P101038 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P101038 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 10667 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 45182 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 13961 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 24287 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 33462 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12148 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48344 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 5347 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 42821 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57585 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 36100 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P101038 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P101038 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321712 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321709 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893520 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893501 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321707 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30893524 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16981652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321710 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10321712 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P101038 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321712 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321718 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321716 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321709 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321719 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321708 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321715 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321707 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321710 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321717 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321720 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10321711 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P101038 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 6296 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321703 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321715 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321695 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321700 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321707 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321712 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321691 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321699 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321705 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321697 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321701 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321689 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10321709 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_031962_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007124 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007132 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007128 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007136 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007137 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007135 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007129 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8007126 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8007134 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8007127 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8007123 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 234679_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11738584_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11738584_PM_x_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006093.hg.1 | ENST00000411528.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006093.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006093.hg.1 | TC17000505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006092.hg.1 | ENST00000411528.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006092.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006092.hg.1 | TC17000505.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012257.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202349.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202349.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202349.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202349.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202349.hg.1 | TC1700012257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | ENST00000377721.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012255.hg.1 | NM_031961.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202347.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202347.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202347.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202347.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202347.hg.1 | TC1700012257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202346.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202346.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202346.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202346.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202346.hg.1 | TC1700012257.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700202348.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202348.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202348.hg.1 | ENST00000411528.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700202348.hg.1 | NM_031962.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700202348.hg.1 | TC1700012257.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 234679_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234679_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_031962_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234679_at6 | - | - |
| Human Genome U133 Set | 234679_PM_at | - | - |
| Human Genome U133 Set | 234679_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009506 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 215439 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_16118241-S | NM_031962.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_16118241-S | AJ406947.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_21762-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006200113 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006200113 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2060402 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_21762 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1693976 | AJ406947.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1693976 | NM_031962.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1693976 | AJ406947.1 | human_transcriptome |