Search
Search
| Gene Symbol: BCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000633.3 | NP_000624.2 | 2 - 3 | 1470 | 81 | |
| XM_047437733.1 | XP_047293689.1 | 1 - 2 | 2012 | 81 | ||
| GenBank mRNA | BC027258.1 | - | 2 - 3 | 977 | 81 | |
| EU287875.1 | - | 1 - 2 | 588 | 81 | ||
| M13994.1 | - | 1 - 2 | 2046 | 81 | ||
| M14745.1 | - | 1 - 2 | 619 | 81 | ||
| X06487.1 | - | 2 - 3 | 1474 | 81 | ||
Set Membership: |
|