Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00608375_m1
          See other DHRS9 GE Assays ›
          Gene Symbol:
          DHRS9
          Gene Name:
          dehydrogenase/reductase 9
          Gene Aliases:
          3-alpha-HSD, 3ALPHA-HSD, RDH-TBE, RDH15, RDHL, RDHTBE, RETSDR8, SDR9C4, hRoDH-E2
          Chromosome Location:
          Chr.2: 169067077 - 169096167 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00608375_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DHRS9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142270.2 NP_001135742.1 3 - 4 700 66
          NM_001142271.2 NP_001135743.1 2 - 3 423 66
          NM_001289763.2 NP_001276692.1 2 - 3 568 66
          NM_001376924.1 NP_001363853.1 2 - 3 532 66
          NM_199204.2 NP_954674.1 3 - 4 833 66
          XM_011510475.2 XP_011508777.1 4 - 5 1045 66
          XM_047442839.1 XP_047298795.1 4 - 5 2725 66
          GenBank mRNA AF067174.1 - 1 - 2 325 66
          AF240697.1 - 4 - 5 415 66
          AF240698.1 - 2 - 3 442 66
          AF295380.1 - 1 - 2 318 66
          AF343729.1 - 2 - 3 471 66
          AF529286.1 - 5 - 6 1821 66
          AF529287.1 - 3 - 4 775 66
          AF529288.1 - 1 - 2 383 66
          AK296625.1 - 2 - 3 568 66
          AY017349.1 - 3 - 4 875 66
          AY359046.1 - 2 - 3 428 66
          BC058883.1 - 2 - 3 428 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009905.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001000.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Human Androgen Panel 1700 Inventoried Human_Androgen Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DHRS9

          Gene Ontology Categories:

          Function(s) Process(es)

          retinoic acid biosynthetic process
          lipid metabolic process
          steroid metabolic process
          androgen metabolic process
          epithelial cell differentiation
          progesterone metabolic process
          retinol metabolic process
          9-cis-retinoic acid biosynthetic process
          alcohol dehydrogenase (NAD) activity
          retinol dehydrogenase activity
          protein binding
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          racemase and epimerase activity
          androsterone dehydrogenase activity
          testosterone dehydrogenase (NAD+) activity
          androstan-3-alpha,17-beta-diol dehydrogenase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000357546.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000412271.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000421653.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000428522.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000432060.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000436483.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000450153.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 ENST00000602501.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001142271.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_001289763.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009905.hg.1 NM_199204.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 116895 - -
          ABI Human Genome Survey Microarray Version 2.0 202451 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-l697b:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline