Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00608644_m1
          See other TIGAR GE Assays ›
          Gene Symbol:
          TIGAR
          Gene Name:
          TP53 induced glycolysis regulatory phosphatase
          Gene Aliases:
          C12orf5, FR2BP
          Chromosome Location:
          Chr.12: 4321213 - 4360028 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00608644_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TIGAR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020375.3 NP_065108.1 3 - 4 251 115
          GenBank mRNA AJ272206.1 - 3 - 4 259 115
          AK313226.1 - 3 - 4 244 115
          AY425618.1 - 3 - 4 230 115
          BC012340.1 - 3 - 4 230 115
          DB480618.1 - 3 - 4 274 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200006557.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12000061.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TIGAR

          Gene Ontology Categories:

          Function(s) Process(es)

          mitophagy
          response to ischemia
          fructose 2,6-bisphosphate metabolic process
          glycolytic process
          autophagy
          apoptotic process
          response to stress
          cellular response to DNA damage stimulus
          response to xenobiotic stimulus
          response to gamma radiation
          cardiac muscle cell apoptotic process
          positive regulation of cardiac muscle cell apoptotic process
          glucose catabolic process to lactate via pyruvate
          regulation of programmed cell death
          negative regulation of programmed cell death
          regulation of pentose-phosphate shunt
          positive regulation of DNA repair
          negative regulation of glycolytic process
          positive regulation of nucleobase-containing compound metabolic process
          positive regulation of phosphate metabolic process
          intestinal epithelial cell development
          cellular response to cobalt ion
          cellular response to hypoxia
          reactive oxygen species metabolic process
          negative regulation of macromitophagy
          regulation of response to DNA damage checkpoint signaling
          positive regulation of hexokinase activity
          negative regulation of glucose catabolic process to lactate via pyruvate
          positive regulation of pentose-phosphate shunt
          negative regulation of reactive oxygen species metabolic process
          catalytic activity
          fructose-2,6-bisphosphate 2-phosphatase activity
          protein binding
          hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000539671.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 ENST00000539671.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000585.hg.1 ENST00000179259.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000585.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000585.hg.1 TC12000061.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12000587.hg.1 ENST00000179259.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000587.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000587.hg.1 TC12000061.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12000590.hg.1 ENST00000179259.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000590.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000590.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000590.hg.1 TC12000061.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12000586.hg.1 ENST00000179259.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000586.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000586.hg.1 TC12000061.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12000589.hg.1 ENST00000179259.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000589.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12000589.hg.1 NM_020375.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12000589.hg.1 TC12000061.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000539671.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 ENST00000539671.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay TC1200006557.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145091.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145091.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145091.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145091.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145091.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145091.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145091.hg.1 TC1200006557.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200145084.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145084.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145084.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145084.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145084.hg.1 TC1200006557.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200145085.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145085.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145085.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145085.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145085.hg.1 TC1200006557.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200145083.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145083.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145083.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145083.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145083.hg.1 TC1200006557.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200145087.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145087.hg.1 ENST00000539671.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145087.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145087.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145087.hg.1 ENST00000539671.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145087.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145087.hg.1 TC1200006557.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200145090.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145090.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145090.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145090.hg.1 ENST00000179259.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145090.hg.1 ENST00000537251.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200145090.hg.1 NM_020375.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200145090.hg.1 TC1200006557.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline