Search
Search
| Gene Symbol: CYBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000101.4 | NP_000092.2 | 1 - 2 | 98 | 110 | |
| XM_011522905.4 | XP_011521207.1 | 1 - 2 | 98 | 110 | ||
| GenBank mRNA | BC006465.1 | - | 1 - 2 | 85 | 110 | |
| BT006861.1 | - | 1 - 2 | 58 | 110 | ||
| BX109994.1 | - | 1 - 2 | 94 | 110 | ||
| M21186.1 | - | 1 - 2 | 86 | 110 | ||
Set Membership: |
|