Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00609523_m1
          See other CHRNB4 GE Assays ›
          Gene Symbol:
          CHRNB4
          Gene Name:
          cholinergic receptor nicotinic beta 4 subunit
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 78623282 - 78655586 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00609523_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CHRNB4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000750.3 NP_000741.1 4 - 5 475 75
          XM_011521186.2 XP_011519488.1 6 - 7 505 75
          XM_011521187.2 XP_011519489.1 5 - 6 412 75
          XM_011521190.2 XP_011519492.1 3 - 4 643 75
          XM_011521191.2 XP_011519493.1 3 - 4 223 75
          XM_017021885.1 XP_016877374.1 4 - 5 570 75
          XM_017021886.1 XP_016877375.1 4 - 5 2377 75
          XM_017021887.1 XP_016877376.1 4 - 5 379 75
          XM_017021888.1 XP_016877377.1 4 - 5 379 75
          XM_017021889.1 XP_016877378.1 4 - 5 379 75
          GenBank mRNA BC096080.1 - 4 - 5 478 40000733 75
          BC096082.1 - 4 - 5 478 40000737 75
          U48861.1 - 4 - 5 483 75
          U62439.1 - 4 - 5 449 75
          X68275.1 - 3 - 4 223 75
          Y08416.1 - 4 - 5 454 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500010897.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001706.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Human Neurotransmitters Human Ligand Gated Ion Channels 1700 Inventoried Human_Ligand_Gated_Ion_Channel Human_Neurotransmitter Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CHRNB4

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          action potential
          ion transport
          smooth muscle contraction
          regulation of smooth muscle contraction
          signal transduction
          synaptic transmission, cholinergic
          locomotory behavior
          response to nicotine
          behavioral response to nicotine
          regulation of neurotransmitter secretion
          protein heterooligomerization
          positive regulation of transmission of nerve impulse
          synaptic transmission involved in micturition
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          drug binding
          ligand-gated ion channel activity
          acetylcholine receptor activity
          acetylcholine binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15016678.hg.1 ENST00000261751.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15016678.hg.1 ENST00000559849.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15016678.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15016678.hg.1 TC15001706.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15016681.hg.1 ENST00000261751.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15016681.hg.1 ENST00000412074.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15016681.hg.1 ENST00000559849.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15016681.hg.1 ENST00000560511.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15016681.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15016681.hg.1 TC15001706.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500185541.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185541.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185541.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay PSR1500185541.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185541.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185541.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay PSR1500185541.hg.1 TC1500010897.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay PSR1500185543.hg.1 TC1500010897.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000261751.7 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000412074.6 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 ENST00000559849.5 Ensembl
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_000750.4 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Affymetrix Human Clariom D Assay TC1500010897.hg.1 NM_001256567.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 163215 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline