Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00610419_g1
          See other AIF1 GE Assays ›
          Gene Symbol:
          AIF1
          Gene Name:
          allograft inflammatory factor 1
          Gene Aliases:
          AIF-1, IBA1, IRT-1, IRT1
          Chromosome Location:
          Chr.6: 31615209 - 31617025 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00610419_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AIF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318970.1 NP_001305899.1 2 - 3 145 92
          NM_001623.4 NP_001614.3 2 - 3 206 92
          XM_005248870.4 XP_005248927.1 2 - 3 198 92
          XM_017010332.1 XP_016865821.1 2 - 3 142 92
          GenBank mRNA AF088038.1 - 2 - 3 152 92
          AK290771.1 - 2 - 3 141 92
          AV716806.1 - 2 - 3 126 92
          AV735626.1 - 2 - 3 252 92
          BC009474.1 - 2 - 3 185 4249306 92
          CB215777.1 - 2 - 3 98 92
          CD521003.1 - 2 - 3 113 92
          D86438.1 - 2 - 3 94 92
          DB502939.1 - 2 - 3 137 92
          EF070983.1 - 2 - 3 85 92
          JQ782414.1 - 2 - 3 180 92
          JQ782415.1 - 2 - 3 119 92
          U19713.1 - 2 - 3 109 92
          U49392.1 - 2 - 3 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007598.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000373.hg.1, TC6_cox_hap2000069.hg.1, TC6_dbb_hap3000060.hg.1, TC6_mcf_hap5000055.hg.1, TC6_qbl_hap6000060.hg.1, TC6_ssto_hap7000056.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the JQ782415.1,NM_001318970.1,XM_017010332.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AIF1

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of protein phosphorylation
          phagocytosis, engulfment
          inflammatory response
          negative regulation of gene expression
          positive regulation of muscle hyperplasia
          Rac protein signal transduction
          actin filament polymerization
          cellular response to extracellular stimulus
          cellular response to hormone stimulus
          positive regulation of T cell proliferation
          negative regulation of apoptotic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of smooth muscle cell proliferation
          negative regulation of smooth muscle cell proliferation
          response to axon injury
          actin filament bundle assembly
          response to glucocorticoid
          response to electrical stimulus
          cellular response to morphine
          cellular response to interferon-gamma
          cellular response to hydroperoxide
          negative regulation of smooth muscle cell chemotaxis
          positive regulation of smooth muscle cell chemotaxis
          positive regulation of monocyte chemotaxis
          ruffle assembly
          positive regulation of G1/S transition of mitotic cell cycle
          positive regulation of T cell migration
          molecular_function
          calcium ion binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5000825.hg.1 TC6_mcf_hap5000055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5000825.hg.1 NM_001623.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5000825.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001010.hg.1 TC6_cox_hap2000069.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001010.hg.1 NM_001623.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001010.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3000953.hg.1 TC6_dbb_hap3000060.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3000953.hg.1 NM_001623.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3000953.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06003627.hg.1 TC06000373.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR06003627.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06003627.hg.1 ENST00000376059.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06003627.hg.1 NM_001623.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06003627.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7000802.hg.1 TC6_ssto_hap7000056.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7000802.hg.1 NM_001623.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7000802.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6000991.hg.1 TC6_qbl_hap6000060.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6000991.hg.1 NM_001623.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6000991.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376049.4 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007598.hg.1 NM_032955.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000337917.11 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000376059.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000466820.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 ENST00000497362.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600151811.hg.1 TC0600007598.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline