Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00611659_s1
          See other FLJ31945 GE Assays ›
          Gene Symbol:
          FLJ31945
          Gene Name:
          uncharacterized LOC440137
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00611659_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ31945
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK056507.1 - 1 1552 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16634843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08843409 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08843417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16634851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08843408 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29291388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16634840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16634850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08843410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08843416 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3322230 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40636 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3322230 - -
          Agilent Human 1B Microarray G4111A A_32_P221218 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556578_PM_a_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1300007198.hg.1 ENST00000618151.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300007198.hg.1 ENST00000618151.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1556578_a_at - -
          Human Genome U133 Plus 2.0 Array 1556578_PM_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556578_a_at8 - -
          Human Genome U95 Set 85220_PM_at - -
          Human Genome U95 Set 85220_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 233385 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1791162 AK056507.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1743256 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1791162 AK056507.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743256 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1743256 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1791162 AK056507.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1791162 AK056507.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1743256 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1743256 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1791162 AK056507.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline