Search
Search

HNF1A-AS1| Gene Symbol: HNF1A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024345.1 | 1 | 1748 | 77 | ||
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654203 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29090157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29090163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29090160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29090156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29090168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16597567 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654196 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654213 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654215 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29090174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654209 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654205 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08654201 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 36136 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 8271 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 704574 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 44184 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P314515 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P314515 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 37216 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P137992 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P137992 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 51206 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P314515 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P314515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654221 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654233 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654220 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654227 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654234 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654225 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654230 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08654221 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P314515 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P137992 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P137992 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P137992 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P314515 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 116879 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 71975 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P137992 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P807070 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654261 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654240 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654222 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16597570 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16597574 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654196 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654224 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16597572 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654199 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16597581 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654248 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654236 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654227 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654238 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654201 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654260 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654233 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654213 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654207 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654246 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654219 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654229 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16597569 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654206 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654203 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654242 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654243 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654250 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654258 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654255 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654244 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654253 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654226 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654209 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08654237 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P137992 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7967108 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7967107 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000537361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000537361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000537361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000619441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | ENST00000619441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026950.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026950.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026950.hg.1 | TC12002057.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200195029.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195029.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195029.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195029.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195029.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195029.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195029.hg.1 | TC1200012188.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195030.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195030.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195030.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195030.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195030.hg.1 | TC1200012188.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195027.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195027.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195027.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195027.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195027.hg.1 | TC1200012188.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195028.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195028.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195028.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195028.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195028.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195028.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195028.hg.1 | TC1200012188.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000433033.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000535301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000537361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000537361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000537361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000539163.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000619441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | ENST00000619441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012188.hg.1 | NR_024345.1 | Refseq |
| Human Genome U95 Set | 87167_at | - | - |
| Human Genome U95 Set | 87167_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048106 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048106 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 231795 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3245684 | NR_024345.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3245684 | AK096009.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3245684 | NR_024345.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3245684 | AK096009.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245684 | NR_024345.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245684 | AK096009.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245684 | NR_024345.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245684 | AK096009.1 | human_transcriptome |