Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00703816_s1
          See other FLJ38773 GE Assays ›
          Gene Symbol:
          FLJ38773
          Gene Name:
          uncharacterized protein FLJ38773
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00703816_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ38773
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK096092.1 - 1 650 70

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 168625 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 16405 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3343412 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P271765 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12383 - -
          Agilent Human Genome CGH Microarray 44B A_14_P121685 - -
          Agilent Human Genome CGH Microarray 44A A_14_P121685 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61520 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P121685 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P271765 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P121685 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P121685 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P121685 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P271765 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 73199 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 49854 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P121685 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P121685 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8065759 - -
          Affymetrix Human Gene 1.1 ST Array 8065758 - -
          Affymetrix HT HG-U133+ PM Array Plate 1559614_PM_at - -
          Human Genome U133 Plus 2.0 Array 1559614_at - -
          Human Genome U133 Plus 2.0 Array 1559614_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) XR_001422_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) XR_001008_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559614_at8 - -
          Human Genome U95 Set 68670_r_at - -
          Human Genome U95 Set 68670_PM_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 318756 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1772626 AK096092.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1772626 AK096092.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1772626 AK096092.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1772626 AK096092.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 4590706 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_38368 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1772626 AK096092.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1772626 AK096092.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1772626 AK096092.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline