Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00706646_s1
          See other FLJ20712 GE Assays ›
          Gene Symbol:
          FLJ20712
          Gene Name:
          uncharacterized FLJ20712
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 33726447 - 33729222 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00706646_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FLJ20712
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017012879.1 XP_016868368.1 1 1499 80
          GenBank mRNA AK000719.1 - 2 547 80
          AK225487.1 - 2 547 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627763 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408740 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627796 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408722 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408744 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15926217 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408768 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408734 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627765 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408756 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408654 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15926208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408675 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15926207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627831 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408739 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408761 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627733 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627757 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627743 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408757 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408720 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627745 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627762 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627731 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627836 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25627838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05408721 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P42684 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P266309 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34281 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12453 - -
          Agilent Human Genome CGH Microarray 44B A_14_P115986 - -
          Agilent Human Genome CGH Microarray 44A A_14_P115986 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 59626 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P42684 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P266309 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P115986 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P115986 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 158862 BC040926.1 genbank_mrna_human
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 97732 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P115986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8132246 - -
          Affymetrix Human Gene 1.0 ST Array 8132247 - -
          Affymetrix Human Gene 1.1 ST Array 8132245 - -
          Affymetrix HT HG-U133+ PM Array Plate 220678_PM_at - -
          Human Genome U133 Plus 2.0 Array 220678_at - -
          Human Genome U133 Plus 2.0 Array 220678_PM_at - -
          Human Genome U133A 2.0 Array 220678_at - -
          Human Genome U133A 2.0 Array 220678_PM_at - -
          AffymetrixPathwork Diagnostics Pathchip pw540093 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 220678_at11 - -
          Human Genome U133 Set 220678_PM_at - -
          Human Genome U133 Set 220678_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 121634 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1779702 AK000719.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1779702 AK225487.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1671345 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1671345 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1779702 AK000719.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1779702 AK225487.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1779702 AK000719.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1779702 AK225487.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline