| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
7912794 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912796 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912792 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912789 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912785 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912797 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912786 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912799 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912798 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912788 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912793 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912791 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912795 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912790 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
7912787 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
7912784 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
219139_PM_s_at |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
215827_PM_x_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000589964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000589964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000589964.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000263511.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000420820.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000589964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000589964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000589964.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000590118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
ENST00000591348.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013045.hg.1 |
NR_023386.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
215827_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
215827_PM_x_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
219139_s_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
219139_PM_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
219139_PM_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
215827_PM_x_at |
- |
- |
| Human Genome U133A 2.0 Array |
219139_s_at |
- |
- |
| Human Genome U133A 2.0 Array |
215827_x_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw529605 |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw592366 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
215827_x_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
219139_s_at9 |
- |
- |
| Human Genome U95 Set |
47492_PM_at |
- |
- |
| Human Genome U95 Set |
47492_at |
- |
- |
| Human Genome U133 Set |
215827_x_at |
- |
- |
| Human Genome U133 Set |
215827_PM_x_at |
- |
- |
| Human Genome U133 Set |
219139_s_at |
- |
- |
| Human Genome U133 Set |
219139_PM_s_at |
- |
- |