Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00736644_m1
          See other SYT16 GE Assays ›
          Gene Symbol:
          SYT16
          Gene Name:
          synaptotagmin 16
          Gene Aliases:
          CHR14SYT, SYT14L, Strep14, syt14r, yt14r
          Chromosome Location:
          Chr.14: 61812156 - 62112825 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00736644_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SYT16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031914.2 NP_114120.2 4 - 5 1634 77
          XM_006720272.3 XP_006720335.1 6 - 7 2094 77
          XM_011537219.1 XP_011535521.1 6 - 7 2171 77
          XM_011537220.1 XP_011535522.1 6 - 7 2171 77
          XM_011537221.2 XP_011535523.1 6 - 7 2171 77
          XM_011537222.2 XP_011535524.1 6 - 7 2171 77
          XM_011537223.2 XP_011535525.1 5 - 6 1208 77
          XM_011537224.2 XP_011535526.1 6 - 7 1354 77
          XM_011537225.2 XP_011535527.1 6 - 7 1653 77
          XM_011537227.2 XP_011535529.2 4 - 5 1031 77
          XM_011537228.2 XP_011535530.1 4 - 5 974 77
          XM_011537229.2 XP_011535531.1 3 - 4 818 77
          XM_017021693.1 XP_016877182.1 6 - 7 2171 77
          XM_017021694.1 XP_016877183.1 6 - 7 1860 77
          XM_017021695.1 XP_016877184.1 7 - 8 2005 77
          XM_017021696.1 XP_016877185.1 5 - 6 1153 77
          XM_017021697.1 XP_016877186.1 5 - 6 1161 77
          XM_017021698.1 XP_016877187.1 5 - 6 1277 77
          GenBank mRNA AB102949.1 - 4 - 5 1377 77
          AJ617628.1 - 4 - 5 1635 77
          BC040924.1 - 2 - 3 526 77
          BC117363.1 - 4 - 5 1493 40125914 77
          BC143598.1 - 4 - 5 1493 9052110 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400007395.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000375.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SYT16

          Gene Ontology Categories:

          Function(s) Process(es)

          vesicle fusion
          regulation of calcium ion-dependent exocytosis
          calcium ion-regulated exocytosis of neurotransmitter
          calcium ion binding
          protein binding
          calcium-dependent phospholipid binding
          syntaxin binding
          clathrin binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14004188.hg.1 ENST00000430451.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14004188.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14004188.hg.1 TC14000375.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14004187.hg.1 ENST00000430451.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14004187.hg.1 NM_031914.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14004187.hg.1 TC14000375.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400009373.hg.1 ENST00000553990.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009373.hg.1 ENST00000553990.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400009373.hg.1 ENST00000553990.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 ENST00000568344.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 ENST00000568344.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400151556.hg.1 TC1400007395.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000430451.3 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000555409.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 ENST00000568344.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400007395.hg.1 NM_031914.2 Refseq
          Affymetrix Human Clariom D Assay TC1400009373.hg.1 ENST00000553990.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009373.hg.1 ENST00000553990.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400009373.hg.1 ENST00000553990.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline