Search
Search

SULT1A1| Gene Symbol: SULT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177536.3 | NP_803880.1 | 2 - 3 | 520 | 128 | |
| GenBank mRNA | AL558375.3 | - | 2 - 3 | 518 | 128 | |
| BI460081.1 | - | 2 - 3 | 483 | 128 | ||
Set Membership: |
|