Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00741306_mH
          See other MSN GE Assays ›
          Gene Symbol:
          MSN
          Gene Name:
          moesin
          Gene Aliases:
          HEL70, IMD50
          Chromosome Location:
          Chr.X: 65588377 - 65741931 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00741306_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MSN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001440778.1 4 - 5 690 128
          NM_002444.3 NP_002435.1 4 - 5 654 128
          XM_005262269.3 XP_005262326.1 4 - 5 690 128
          XM_011530959.1 XP_011529261.1 4 - 5 580 128
          XM_047442129.1 XP_047298085.1 4 - 5 5168 128
          XM_047442130.1 XP_047298086.1 5 - 6 783 128
          XM_047442131.1 XP_047298087.1 5 - 6 4508 128
          GenBank mRNA BC001112.2 - 4 - 5 636 128
          BC011827.2 - 4 - 5 636 128
          BC017293.2 - 4 - 5 617 128
          EU794636.1 - 4 - 5 617 128
          M69066.1 - 4 - 5 566 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0X00011302.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0X000353.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MSN

          Gene Ontology Categories:

          Function(s) Process(es)

          immunological synapse formation
          cytoskeleton organization
          leukocyte cell-cell adhesion
          regulation of cell shape
          regulation of cell size
          positive regulation of gene expression
          gland morphogenesis
          membrane to membrane docking
          T cell proliferation
          establishment of epithelial cell apical/basal polarity
          positive regulation of protein catabolic process
          leukocyte migration
          establishment of endothelial barrier
          T cell aggregation
          positive regulation of podosome assembly
          T cell migration
          regulation of organelle assembly
          positive regulation of protein localization to early endosome
          regulation of lymphocyte migration
          positive regulation of early endosome to late endosome transport
          double-stranded RNA binding
          actin binding
          receptor binding
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          enzyme binding
          protein kinase binding
          cell adhesion molecule binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR0X004551.hg.1 ENST00000360270.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X004551.hg.1 NM_002444.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X004551.hg.1 TC0X000353.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0X004550.hg.1 ENST00000360270.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0X004550.hg.1 NM_002444.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0X004550.hg.1 TC0X000353.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0X00177614.hg.1 ENST00000360270.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177614.hg.1 NM_002444.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177614.hg.1 ENST00000360270.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177614.hg.1 NM_002444.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177614.hg.1 TC0X00011302.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0X00177615.hg.1 ENST00000360270.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177615.hg.1 NM_002444.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177615.hg.1 ENST00000360270.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0X00177615.hg.1 NM_002444.2 Refseq
          Affymetrix Human Clariom D Assay PSR0X00177615.hg.1 TC0X00011302.hg.1 Clariom D TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline