Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00745096_s1
          See other KLHL30-AS1 GE Assays ›
          Gene Symbol:
          KLHL30-AS1
          Gene Name:
          KLHL30 antisense RNA 1
          Gene Aliases:
          C2orf19
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00745096_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KLHL30-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC031945.1 - 1 2950 103

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 164390 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 723159 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 6322 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P941006 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 22499 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00627186 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P941006 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P00627186 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991616 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991597 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991634 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991581 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991606 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991583 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991590 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991624 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991627 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991619 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991629 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991589 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991632 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991636 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991608 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991598 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991576 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991599 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991610 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991574 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991600 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991630 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991620 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991584 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991604 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991618 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991587 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991594 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991612 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991573 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991578 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991602 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P01991593 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P01991637 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398372 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398369 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398366 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P01991638 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398354 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398367 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398365 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398370 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398371 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P01991639 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398352 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398364 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398368 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15398353 - -
          Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P01991601 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00627186 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P941006 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 10928 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 72161 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00627186 - -
          Agilent Human 1B Microarray G4111A A_32_P476182 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00627186 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P16092248 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8059966 - -
          Affymetrix Human Gene 1.0 ST Array 8059967 - -
          Affymetrix Human Gene 1.0 ST Array 8059968 - -
          Affymetrix Human Gene 1.1 ST Array 8059965 - -
          Affymetrix HT HG-U133+ PM Array Plate 1554782_PM_at - -
          Human Genome U133 Plus 2.0 Array 1554782_PM_at - -
          Human Genome U133 Plus 2.0 Array 1554782_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NM_001024676_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1554782_at9 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 233925 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1738410 BC031945.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1738410 BC031945.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1738410 BC031945.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_16989-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1738410 BC031945.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1738410 BC031945.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 5560309 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_16989 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1738410 BC031945.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1738410 BC031945.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1738410 BC031945.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline