Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00746672_s1
          See other DPY19L2P4 GE Assays ›
          Gene Symbol:
          DPY19L2P4
          Gene Name:
          DPY19L2 pseudogene 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 90119400 - 90125600 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00746672_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DPY19L2P4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003551.1 1 202 152
          GenBank mRNA AK098759.1 - 1 181 153
          BC031277.1 - 1 158 153
          BC047471.1 - 1 148 153
          BX537815.1 - 1 202 152
          DQ287934.1 - 1 202 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700008290.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000535.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK098759.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 AB027471.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 AB075513.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 AB209681.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 AK313084.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 BC024004.2 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 BC040183.2 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 NM_001100397.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 NM_007023.3 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CGH Microarray 4x180K 120716 U78516.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 746993 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 141764 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 AB006198.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 AF353625.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 BC001058.2 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 BC018607.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 BT006637.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 BX648500.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 CR611735.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 CR626621.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 NM_005146.4 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 172499 Y14314.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 345783 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 119480 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 40142 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P16168669 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01751636 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P177948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25853664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15987314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25853638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05602811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15987314 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25853638 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25853664 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05602811 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05602816 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05602811 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P16171845 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P16168669 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003551_at - -
          Affymetrix Human Gene 1.0 ST Array 8134028 - -
          Affymetrix Human Gene 1.0 ST Array 8134027 - -
          Affymetrix Human Gene 1.0 ST Array 8134029 - -
          Affymetrix Human Gene 1.1 ST Array 8134026 - -
          Affymetrix Human Transcriptome Array 2.0 PSR07007590.hg.1 TC07000535.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07007590.hg.1 ENST00000497063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07007590.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07007592.hg.1 TC07000535.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07007592.hg.1 ENST00000497063.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07007592.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157596.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157596.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157596.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157596.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157596.hg.1 TC0700008290.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700157586.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157586.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157586.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157586.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157586.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157586.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157586.hg.1 TC0700008290.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700157594.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157594.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157594.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157594.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157594.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157594.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157594.hg.1 TC0700008290.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700157585.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157585.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157585.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157585.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157585.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157585.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157585.hg.1 TC0700008290.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay TC0700008290.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157583.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157583.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157583.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157583.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157583.hg.1 TC0700008290.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700157597.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157597.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157597.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157597.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157597.hg.1 TC0700008290.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700157584.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157584.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157584.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157584.hg.1 ENST00000479881.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157584.hg.1 ENST00000497063.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700157584.hg.1 NR_003551.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700157584.hg.1 TC0700008290.hg.1 Clariom D TC
          Human Genome U133 Plus 2.0 Array 1557290_at - -
          Human Genome U133 Plus 2.0 Array 1557290_PM_at - -
          Human Genome U95 Set 63046_PM_at - -
          Human Genome U95 Set 62929_PM_at - -
          Human Genome U95 Set 63046_at - -
          Human Genome U95 Set 62929_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034680 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034681 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034681 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A034680 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 229935 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1741908 BC047471.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1741908 BX537815.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1741908 DQ287934.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1741908 NR_003551.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1741908 BC031277.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1741908 BC047471.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1741908 BX537815.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1741908 DQ287934.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1741908 NR_003551.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1741908 BC031277.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1741908 BC047471.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1741908 BX537815.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1741908 DQ287934.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1741908 NR_003551.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1741908 BC031277.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1741908 BC047471.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1741908 BX537815.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1741908 DQ287934.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1741908 NR_003551.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1741908 BC031277.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline