| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151465.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151465.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151465.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151465.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151465.hg.1 |
TC0X00007516.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007516.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151463.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151463.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151463.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151463.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151463.hg.1 |
TC0X00007516.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00151467.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151467.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151467.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151467.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151467.hg.1 |
TC0X00007516.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00151466.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151466.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151466.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151466.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151466.hg.1 |
TC0X00007516.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00151464.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151464.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151464.hg.1 |
ENST00000399711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00151464.hg.1 |
NR_103715.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00151464.hg.1 |
TC0X00007516.hg.1 |
Clariom D TC |