| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074332 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074290 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074334 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207765 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074340 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207825 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074322 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15415511 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074303 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207796 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15415498 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074284 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074299 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207779 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074347 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074345 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074307 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207749 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074324 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207788 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074329 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207761 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074343 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074293 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207782 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074344 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074288 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074319 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074346 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074335 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074289 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074349 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074325 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074337 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074281 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074283 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207789 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074291 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074294 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074292 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207742 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074350 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207803 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074316 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074358 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15415499 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074352 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074295 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207762 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074286 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074333 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074308 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074317 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207814 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074328 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074306 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207740 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207778 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15415493 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207812 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074315 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074336 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207795 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15415494 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15415495 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15415491 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074323 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P22207766 |
- |
- |
| Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02074353 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
542427 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
1291 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
933109 |
- |
- |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
514658 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
410433 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
4317 |
- |
- |
| Agilent SurePrint G3 Human CNV Microarray 2x400K |
90451 |
- |
- |
| Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3507384 |
- |
- |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
62825 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P00653398 |
- |
- |
| Agilent Human Genome CGH + SNP Microarray |
A_16_P00653398 |
- |
- |
| SurePrint G3 Human GE Microarray G4851A |
A_33_P3507384 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00653398 |
- |
- |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00653409 |
- |
- |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120648 |
AB033110.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120648 |
AK123320.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120648 |
AK291481.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120648 |
BC051698.1 |
genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
120648 |
NM_015693.2 |
refseq_mrna_human_NM |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00653407 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00653415 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00653398 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00653409 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P36175989 |
- |
- |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00653405 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00653409 |
- |
- |
| Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00653398 |
- |
- |