Search
Search

PSMC2| Gene Symbol: PSMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204453.1 | NP_001191382.1 | 1 - 2 | 254 | 118 | |
| NM_002803.4 | NP_002794.1 | 1 - 2 | 135 | 118 | ||
| GenBank mRNA | AB075520.1 | - | 1 - 2 | 115 | 118 | |
| AK298529.1 | - | 1 - 2 | 102 | 118 | ||
| AK298821.1 | - | 1 - 2 | 115 | 118 | ||
| AK312648.1 | - | 1 - 2 | 106 | 118 | ||
| BC002589.2 | - | 1 - 2 | 98 | 118 | ||
| BE269755.1 | - | 2 - 3 | 331 | 118 | ||
| BM754312.1 | - | 1 - 2 | 136 | 118 | ||
| D11094.1 | - | 1 - 2 | 132 | 118 | ||
| HM005610.1 | - | 1 - 2 | 136 | 118 | ||
Set Membership: |
|