| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000366.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000366.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000366.hg.1 |
TC14002288.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000367.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000367.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000367.hg.1 |
TC14002288.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000369.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000369.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14000369.hg.1 |
TC14002288.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006540.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144640.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144640.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144640.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144640.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144640.hg.1 |
TC1400006540.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400144641.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144641.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144641.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144641.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144641.hg.1 |
TC1400006540.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400144643.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144643.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144643.hg.1 |
ENST00000304625.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400144643.hg.1 |
NM_002934.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400144643.hg.1 |
TC1400006540.hg.1 |
Clariom D TC |