| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016174.hg.1 |
ENST00000446824.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016174.hg.1 |
NM_021018.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016174.hg.1 |
TC06001353.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011140.hg.1 |
ENST00000618052.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011140.hg.1 |
NM_021018.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011140.hg.1 |
NM_021018.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011140.hg.1 |
ENST00000618052.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011140.hg.1 |
NM_021018.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011140.hg.1 |
NM_021018.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175359.hg.1 |
ENST00000618052.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175359.hg.1 |
NM_021018.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175359.hg.1 |
ENST00000618052.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600175359.hg.1 |
NM_021018.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600175359.hg.1 |
TC0600011140.hg.1 |
Clariom D TC |